Skip to Content
Merck
All Photos(1)

Key Documents

OP180

Sigma-Aldrich

Anti-Noxa Mouse mAb (114C307)

liquid, clone 114C307, Calbiochem®

Synonym(s):

Anti-Noxa Antibody, Mouse Anti-Noxa, Noxa Detection Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

114C307, monoclonal

form

liquid

contains

≤0.1% sodium azide as preservative

species reactivity

human

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

isotype

IgG1

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PMAIP1(5366)

General description

Purified mouse monoclonal antibody. Recognizes the ~11 kDa Noxa protein.
Recognizes the ~11 kDa Noxa protein in RL-7 cells.
This Anti-Noxa Mouse mAb (114C307) is validated for use in Immunoblotting for the detection of Noxa.

Immunogen

Human
a recombinant protein consisting of human Noxa fused to GST

Application


Immunoblotting (1-2 g/ml, chemiluminescence)

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Standard Handling (A)

Physical form

In PBS, 0.05% BSA.

Reconstitution

Following initial thaw, aliquot and freeze (-20°C;)

Analysis Note

Negative Control
Normal mouse serum
Positive Control
RL-7 cell lysate

Other Notes

Antibody should be titrated for optimal results in individual system.
Oda, E., et al. 2000. Science288, 1053.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nabanita Mukherjee et al.
Cancers, 12(8) (2020-08-09)
There is an urgent need to develop treatments for patients with melanoma who are refractory to or ineligible for immune checkpoint blockade, including patients who lack BRAF-V600E/K mutations. This is often the case in patients diagnosed with rare melanoma subtypes
Tao Hu et al.
Molecular carcinogenesis, 58(1), 42-54 (2018-09-06)
Increasing evidence suggests that deubiquitinase USP7 participates in tumor progression by various mechanisms and serves as a potential therapeutic target. However, its expression and role in esophageal cancer remains elusive; the anti-cancer effect by targeting USP7 still needs to be
Santi Rello-Varona et al.
Scientific reports, 9(1), 3816-3816 (2019-03-09)
Soft-tissue sarcomas (STS) are an uncommon and heterogeneous group of malignancies that result in high mortality. Metastatic STS have very bad prognosis due to the lack of effective treatments. Dinaciclib is a model drug for the family of CDK inhibitors.
David J Mallick et al.
Cancers, 12(8) (2020-08-23)
Anti-apoptotic BCL2 proteins are important targets for cancer therapy as cancers depend on their activity for survival. Direct inhibitors of MCL1 have entered clinical trials, although their efficacy may be limited by toxicity. An alternative approach may be to induce
Xinying Li et al.
Oncology letters, 17(2), 1851-1858 (2019-01-25)
Esophageal squamous cell carcinoma (ESCC) is a dominant histological subtype of esophageal cancer with notably high incidence and mortality rates. Pemetrexed is a clinical antifolate therapeutic agent with anticancer properties. The present study aimed to understand whether pemetrexed is able

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service