Skip to Content
Merck
All Photos(1)

Key Documents

NI01

Sigma-Aldrich

Normal Rabbit IgG

This Normal Rabbit IgG is validated for use as a negative control in parallel with specific primary antibodies in ELISA, FC, Immunoblotting, IF, IHC, IP.

Synonym(s):

Normal IgG, Rabbit IgG control, Rabbit immunoglobulin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.43

biological source

rabbit

Quality Level

antibody form

serum

form

liquid

contains

≤0.1% sodium azide as preservative

manufacturer/tradename

Calbiochem®

storage condition

do not freeze

shipped in

wet ice

storage temp.

2-8°C

General description

Native IgG from rabbit serum. IgG is a glycoprotein found in serum and tissue fluids. For use as a negative control in parallel with specific rabbit IgG-type primary antibodies.
This Normal Rabbit IgG is validated for use as a negative control in parallel with specific primary antibodies in ELISA, FC, Immunoblotting, IF, IHC, IP.

Application

For use as a negative control in parallel with specific primary antibodies in:

ELISA

Flow Cytometry

Immunoblotting

Immunofluorescence

Immunohistochemistry

Immunoprecipitation

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Standard Handling (A)

Physical form

In 0.01 M sodium phosphate, 0.25 M NaCl, pH 7.6.

Other Notes

For use as a negative control, perform experiments in parallel using primary antibody or normal serum at the same concentration. A signal obtained with the normal serum indicates a nonspecific interaction; any additional signal with the primary antibody indicates a specific antibody-antigen interaction.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Penghua You et al.
Journal of cellular physiology, 235(5), 4455-4465 (2019-10-23)
Lin28a has been found to enhance glucose uptake and insulin sensitivity. Lin28a alleviates cardiac dysfunction under various pathological conditions. However, the effects and underlying mechanisms of Lin28a on diabetic cardiomyopathy (DCM) are not well-understood. The aim of this study was
Jingnan Pi et al.
Cancer research, 81(10), 2651-2665 (2020-08-14)
N6-methyladenosine (m6A) is the most prevalent internal RNA modification in mammals that regulates homeostasis and function of modified RNA transcripts. Here, we aimed to investigate the role of YTH m6A RNA-binding protein 1 (YTHDF1), a key regulator of m6A methylation
Kyathanahalli S Janardhan et al.
Toxicologic pathology, 47(5), 577-584 (2019-05-09)
The majority of the tumors in the gastrointestinal (GI) tract of rats and mice, with spindle cell morphology, are diagnosed as smooth muscle tumors (SMTs). Similarly, several decades ago human GI tumors with spindle cell morphology were also diagnosed as
Chutchai Piewbang et al.
PloS one, 15(10), e0241212-e0241212 (2020-10-24)
Hepatitis B virus (HBV) is a human pathogen of global concern, while a high diversity of viruses related to HBV have been discovered in other animals during the last decade. Recently, the novel mammalian hepadnavirus, tentatively named domestic cat hepadnavirus
Suruchi Shrestha et al.
Oncogene, 39(36), 5855-5866 (2020-07-04)
Epidermal squamous cell carcinoma (SCC) is a common and highly invasive form of cancer. SCC arises due to ultraviolet light exposure and is associated with increased expression of pro-cancer genes and reduced expression of cancer suppressors. Actin-Like Protein 6A (ACTL6A

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service