Skip to Content
Merck
All Photos(1)

Key Documents

MXPESPWEL

Millipore

MicropreSure® Sanitary Sampling Valve

valve size 1/4 in., 316L stainless steel

Synonym(s):

Stainless steel sampling valve

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116129
eCl@ss:
32160606
NACRES:
NC.57

material

316L stainless steel

packaging

pkg of 1 ea

manufacturer/tradename

MicropreSURE®

valve size

1/4 in.

application(s)

bioburden testing
food and beverages
pharmaceutical

shipped in

ambient

General description

MicropreSure® sanitary sampling valve is an accessory of the MicropreSure® sampler. MicropreSure® sampler is an on-line filtration sampler to simplify in-process microbiological testing. It is a simple, protective design that allows samples to be aseptically processed right at the collection site thereby, reducing contamination risk. The Sanitary sampling valve is installed at the sampling port outlet having the weld end connection.

Application

MicropreSure® sanitary sampling valve is used for on-line sampling and filtration of aqueous liquids such as beer, dairy, bottled water, pharmaceutical products analysis, soft drink, cider, sports drink, and wine for bioburden testing.

Legal Information

MICROPRESURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service