Skip to Content
Merck
All Photos(3)

Key Documents

MABF2033M

Sigma-Aldrich

Anti-CD31/PECAM-1.3 Antibody, clone MBC 78.3

clone MBC 78.3, from mouse

Synonym(s):

Platelet endothelial cell adhesion molecule, PECAM-1, EndoCAM, GPIIA′, PECA1, CD31

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.43

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

MBC 78.3, monoclonal

species reactivity

human, canine

packaging

antibody small pack of 25 μg

technique(s)

flow cytometry: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

human ... PECAM1(5175)

Related Categories

General description

Platelet endothelial cell adhesion molecule (UniProt: P16284; also known as PECAM-1, EndoCAM. GPIIA′, PECA1, CD31) is encoded by the PECAM1 gene (Gene ID: 5175) in human. PECAM-1 is a single-pass type I membrane protein that is synthesized with a signal peptide (aa 1-27) that is subsequently cleaved to generate the mature protein with a large extracellular region (aa 28-601) that contains six Ig-like C2-type domains, followed by a transmembrane segment (aa 602-620) and a cytoplasmic tail (aa. 621-738 of isoform Long) that plays an essential role in PECAM-1-mediated cellular signaling. PECAM-1 is essential for leukocyte transendothelial migration (TEM) under most inflammatory conditions. It is expressed on platelets and leukocytes and is primarily concentrated at the borders between endothelial cells. Six isoforms of PECAM-1 have been described that are produced by alternative splicing. Isoform Long predominates in all tissues examined, whereas Delta 12 isoform is detected only in trachea and isoform Delta14-15 are only detected in lung tissue. Isoform Delta14 is detected in all tissues examined with the strongest expression in heart. Isoform Delta15 is expressed in brain, testis, ovary, cell surface of platelets, human umbilical vein endothelial cells (HUVECs), Jurkat T-cell leukemia, human erythroleukemia and U-937 histiocytic lymphoma cell lines. Tyrosine 690 residue plays a critical role in TEM and is required for efficient trafficking of PECAM-1 to and from the lateral border recycling compartment (LBRC) and is also required for the LBRC membrane to be targeted around migrating leukocytes. PECAM-1 also prevents phagocyte ingestion of closely apposed viable cells by transmitting ′detachment′ signals, and changes function on apoptosis, promoting tethering of dying cells to phagocytes.

Specificity

Clone MBC 78.3 detects CD31/PECAM-1 in human and canine cells. It targets an epitope within the IgG homology domain 1.

Immunogen

Epitope: extracellular domain
PECAM-1 purified from human platelets.

Application

Anti-CD31/PECAM-1.3, clone MBC 78.3 , Cat. No. MABF2033 is a mouse monoclonal antibody that detects PECAM1 and has been tested for use in Flow Cytometry, Immunocytochemistry, Immunoprecipitation, Agonist or Inhibitor function, and Western Blotting.
Flow Cytometry Analysis: A representative lot detected CD31/PECAM-1.3 in Flow Cytometry applications (Newton-Nash, D.K., et. al. (1999). J Immunol. 163(2):682-8).

Flow Cytometry Analysis: 1 µg from a representative lot detected CD31/PECAM-1.3 in THP-1 cells.

Western Blotting Analysis: A representative lot detected CD31/PECAM-1.3 in Western Blotting applications (Zhao, T., et. al. (2001). J Cell Biol. 152(1):65-73).

Agonist or Inhibitor Analysis: A representative performed agonist or Inhibitor function (Newton-Nash, D.K., et. al. (1999). J Immunol. 163(2):682-8; Reedguist, K.A., et. al. (2000). J Cell Biol. 148(6):1151-8).

Immunoprecipitation Analysis: A representative lot detected CD31/PECAM-1.3 in Immunoprecipitation applications (Newman, D.K., et. al. (2016). Sci Signal. 9(418):ra27).

Immunocytochemistry Analysis: A representative lot detected CD31/PECAM-1.3 in Immunocytochemistry applications (Newman, D.K., et. al. (2016). Sci Signal. 9(418):ra27; Zhao, T., et. al. (2001). J Cell Biol. 152(1):65-73).
Research Category
Inflammation & Immunology

Quality

Evaluated by Immunocytochemistry in un-permeablilized THP-1 cells.

Immunocytochemistry Analysis: A 1:500 dilution of this antibody detected CD31/PECAM-1.3 in un-permeablilized THP-1 cells.

Target description

82.54 kDa calculated.

Physical form

Format: Purified
Protein G purified
Purified mouse monoclonal antibody IgG1 in PBS without azide.

Storage and Stability

Stable for 1 year at -20°C from date of receipt.

Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service