Skip to Content
Merck
All Photos(2)

Key Documents

MAB5404

Sigma-Aldrich

Anti-Nitrotyrosine Antibody, clone 2A8.2

clone 2A8.2, Chemicon®, from mouse

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

2A8.2, monoclonal

species reactivity (predicted by homology)

all

manufacturer/tradename

Chemicon®

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG2b

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... NOS1(4842)

General description

The cellular production of highly reactive nitrogen species derived from nitric oxide, such as peroxynitrite, nitrogen dioxide and nitryl chloride, leads to the nitration of tyrosine resides in tissue proteins. The extent of protein nitrotyrosine formation provides an index of the production of reactive nitrogen species and potential cell damage over a period of time. Nitrotyrosine can be measured by amino-acid analysis of protein hydrolysates and detected, estimated semi-quantitatively and located in cells and tissues by immunocytochemical techniques using antibodies directed against the nitrotyrosine hapten.

Specificity

Reacts with nitrotyrosine.

Immunogen

Nitrated KLH.

Application

Detect Nitrotyrosine using this Anti-Nitrotyrosine Antibody, clone 2A8.2 validated for use in IH(P) & WB.
Immunohistochemistry:
1:50-1:100 on paraffin embedded of a previous lot, 10% neutral buffer formalin fixed mouse liver (animal treated with Zymosan) and normal human cerebellum tissues. The tissues must be pretreated with heat induced epitope retrieval (HIER). Incubation with the MAB5404 was for 10 minutes and the reaction was developed with the Chemicon IHC-Select, HRP-DAB kit (Catalog number DAB150).

Optimal working dilutions must be determined by end user.
Research Category
Neuroscience
Research Sub Category
Oxidative Stress

Quality

Evaluated by Western Blot on Nitrotyrosine Immunoblot Controls.

Western Blot Analysis:
1:500 dilution of this antibody detected Nitrotyrosine on 10 μg of Nitrotyrosine Immunoblot Controls (cat. # 12-354).

Physical form

Format: Purified
Protein A purified
Purified mouse monoclonal IgG2b liquid in buffer containing 0.1% sodium azide.

Storage and Stability

Stable for 1 year at 2-8ºC from date of receipt.

Analysis Note

Control
Nitrated BSA

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ben C Creelan et al.
OncoTargets and therapy, 10, 4239-4250 (2017-09-19)
Omaveloxolone is a semisynthetic oleanane triterpenoid that potently activates Nrf2 with subsequent antioxidant function. We conducted a first-in-human Phase I clinical trial (NCT02029729) with the primary objectives to determine the appropriate dose for Phase II studies, characterize pharmacokinetic and pharmacodynamic
A Kiss et al.
British journal of pharmacology, 155(7), 1015-1024 (2008-10-11)
Exogenous peroxynitrite from nanomolar to micromolar concentrations exerts cardioprotection. Here, we have assessed its effects on ischaemia- and reperfusion-induced ventricular arrhythmias in vivo and a possible role for mitochondrial K(ATP) channels in these effects, using the channel inhibitor 5-hydroxydecanoate (5-HD).
Satoru Hasegawa et al.
Redox report : communications in free radical research, 22(1), 26-34 (2016-02-13)
Duchenne muscular dystrophy (DMD) is a devastating muscle disease caused by a mutation in DMD encoding dystrophin. Oxidative stress accounts for dystrophic muscle pathologies in DMD. We examined the effects of molecular hydrogen in mdx mice, a model animal for
G L Rodríguez-González et al.
International journal of obesity (2005), 39(4), 549-556 (2014-12-17)
Increasing evidence exists that maternal obesity (MO) and overnutrition during pregnancy and lactation have long-lasting consequences for progeny metabolism, cardiovascular and endocrine function. Data on effects of MO on offspring reproduction are limited. We hypothesized that MO during pregnancy and
Guadalupe L Rodríguez-González et al.
The Journal of physiology, 597(23), 5549-5563 (2019-10-09)
Maternal obesity predisposes to metabolic dysfunction in male and female offspring Maternal high-fat diet consumption prior to and throughout pregnancy and lactation accelerates offspring metabolic ageing in a sex-dependent manner This study provides evidence for programming-ageing interactions ABSTRACT: Human epidemiological

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service