Skip to Content
Merck
All Photos(2)

Key Documents

MAB3101

Sigma-Aldrich

Anti-Connexin 45 Antibody, near CT, cytoplasmic, clone 5B9.2

clone 5B9.2, Chemicon®, from mouse

Synonym(s):

Cx45

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

5B9.2, monoclonal

species reactivity

rat, rabbit, human

manufacturer/tradename

Chemicon®

technique(s)

immunocytochemistry: suitable
western blot: suitable

isotype

IgG1

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... GJC1(10052)
rat ... Gjc1(266706)

Specificity

Recognizes Human Connexin 45.

SPECIES REACTIVITIES:

Due to sequence homologies, this antibody is also expected to react with canine (100% homology) and mouse (92% homology) Connexin 45. Also confirmed to work on rat C6 cells. Others not tested.

Immunogen

Epitope: near C-terminus, cytoplasmic
The peptide sequence used for the immunogen was H-Gln-Ala-Tyr-Ser-His-Gln-Asn-Asn-Pro-His-Gly-Pro-Arg-Glu-Cys-OH conjugated to KLH, corresponding to amino acids 354-367 of human connexin 45.

Application

Anti-Connexin 45 Antibody, near C-terminus, cytoplasmic, clone 5B9.2 detects level of Connexin 45 & has been published & validated for use in IC & WB.
Immunblotting (~46 kDa)

Immunocytochemistry

Optimal working dilutions must be determined by end user.
Research Category
Neuroscience
Research Sub Category
Ion Channels & Transporters

Physical form

Format: Purified
Liquid at 1 mg/mL in 0.02M PB with 0.25 M NaCl, pH=7.6 containing 0.1% sodium azide.

Storage and Stability

Maintain at 2-8°C in undiluted aliquots for up to 12 months.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Axonal synapses utilize multiple synaptic ribbons in the mammalian retina.
Kim, HL; Jeon, JH; Koo, TH; Lee, UY; Jeong, E; Chun, MH; Moon, JI; Massey, SC; Kim, IB
Testing null
Hideo Hoshi et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 29(28), 8875-8883 (2009-07-17)
The vertebrate retina is a distinctly laminar structure. Functionally, the inner plexiform layer, in which bipolar cells synapse onto amacrine and ganglion cells, is subdivided into two sublaminae. Cells that depolarize at light offset ramify in sublamina a; those that
Richard Fischer et al.
Gastroenterology, 128(2), 433-448 (2005-02-03)
Gap junctional communication was studied in quiescent and activated hepatic stellate cells. Connexin expression and intercellular dye transfer were studied in rat hepatic stellate cells in culture and in vivo. Protein expression of connexin 43 was up-regulated in activated hepatic
J I Nagy et al.
The European journal of neuroscience, 46(2), 1790-1804 (2017-06-01)
Electrical synapses formed by connexin36 (Cx36)-containing gap junctions between interneurons in the cerebellar cortex have been well characterized, including those formed between basket cells and between Golgi cells, and there is gene reporter-based evidence for the expression of connexin45 (Cx45)
Maria D Mayan et al.
The American journal of pathology, 182(4), 1337-1346 (2013-02-19)
Osteoarthritis (OA) is the most common joint disease and involves progressive degeneration of articular cartilage. The aim of this study was to investigate if chondrocytes from human articular cartilage express gap junction proteins called connexins (Cxs). We show that human

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service