Skip to Content
Merck
All Photos(1)

Key Documents

AP309P

Sigma-Aldrich

Goat Anti-Human IgG Antibody, HRP conjugate

1.0 mg/mL, Chemicon®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.46

biological source

goat

Quality Level

conjugate

peroxidase conjugate

antibody form

affinity purified immunoglobulin

antibody product type

secondary antibodies

clone

polyclonal

form

lyophilized

species reactivity

human

manufacturer/tradename

Chemicon®

concentration

1.0 mg/mL

technique(s)

ELISA: suitable
western blot: suitable

isotype

IgG

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

General description

Immunoglobulin G (IgG) is the main type of glycoprotein antibody found in human serum, secreted by the B cells. It is found in blood and comprises four polypeptide chains, made of two identical 50 kDa γ heavy (H) chains and two identical 25 kDa κ or λ light (L) chains. These chains are inter-linked by di-sulfide bonds. IgG exists in four isotypes IgG1, IgG2, IgG3 and IgG4. IgG has the longest serum half-life compared to other immunoglobulins.

Specificity

Specific for human IgG gamma chain. The cross-reactivities of anti-human IgG (gamma) antibody are tested in an ELISA. Minimal cross-reactivity to light chains.

The reagent is an affinity purified antibody from goat. The purified antibody is conjugated to horseradish peroxidase (HRP) and stabilized in buffer.

Application

ELISA: 1:3,000

Immunohistochemistry: 1:200-1:500 (1-4)

Immunocytochemistry: 1:200-1:500 (1-4)

Western Blot, chromogenic: 1:200-1-500

Western Blot, chemiluminescent: 1:2,000-1:5,000

Optimal working dilutions must be determined by the end user.

Biochem/physiol Actions

Immunoglobulin G (IgG) initiates the classical pathway of the complement system. It neutralizes virus particles and toxins. IgG plays a vital role in antibody-dependent cell-mediated cytotoxicity (ADCC) and might be associated with allergy. Maternal IgG is transferred to the fetus through the placenta that is crucial for the immune defense of the neonate against infections. Digestion of IgG by papain results in the production of fragment antigen-binding (Fab). Pepsin digestion of IgG leads to the generation of fragment crystallizable (Fc). Levels of IgG in severe acute respiratory syndrome coronavirus (SARS-CoV)-2 infected patients can be used as a potential tool to evaluate the severity and prognosis of coronavirus disease 2019 (COVID-19).

Physical form

ImmunoAffinity Purified

0.01M PBS, PH7.1, with 15mg/mL BSA

Storage and Stability

The undiluted antibody is stable at 2-8°C for 12 months. Do not store in a diluted format. DO NOT FREEZE.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Serum polyethylene glycol-specific IgE and IgG in patients with hypersensitivity to COVID-19 mRNA vaccines.
Mouri, et al.
Allergology International, 71, 512-519 (2022)
Alexander A Dolskiy et al.
Frontiers in molecular biosciences, 8, 821506-821506 (2022-02-05)
The novel coronavirus disease COVID-19 has become one of the most socially significant infections. One of the main models for COVID-19 pathogenesis study and anti-COVID-19 drug development is laboratory animals sensitive to the virus. Herein, we report SARS-CoV-2 infection in
Wajihul Hasan Khan et al.
International journal of biological macromolecules, 200, 428-437 (2022-01-21)
Nucleocapsid protein (N protein) is the primary antigen of the virus for development of sensitive diagnostic assays of COVID-19. In this paper, we demonstrate the significant impact of dimerization of the severe acute respiratory syndrome coronavirus 2 (SARS-CoV-2) N-protein on
Xiaotian Tang et al.
Cell reports, 41(8), 111673-111673 (2022-11-24)
In North America, the Lyme disease agent, Borrelia burgdorferi, is commonly transmitted by the black-legged tick, Ixodes scapularis. Tick saliva facilitates blood feeding and enhances pathogen survival and transmission. Here, we demonstrate that I. scapularis complement C1q-like protein 3 (IsC1ql3), a
Marti Cabanes-Creus et al.
Nature communications, 15(1), 1876-1876 (2024-03-15)
Developing clinically predictive model systems for evaluating gene transfer and gene editing technologies has become increasingly important in the era of personalized medicine. Liver-directed gene therapies present a unique challenge due to the complexity of the human liver. In this

Related Content

Products and support for mpox research, offering world-class reagents for immunoassay and molecular assay development.

Products and support for mpox research, offering world-class reagents for immunoassay and molecular assay development.

Products and support for mpox research, offering world-class reagents for immunoassay and molecular assay development.

Products and support for mpox research, offering world-class reagents for immunoassay and molecular assay development.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service