AB5909
Anti-Galanin Antibody
serum, Chemicon®
Synonym(s):
Anti-ETL8, Anti-GAL-GMAP, Anti-GALN, Anti-GLNN, Anti-GMAP
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
biological source
rabbit
Quality Level
antibody form
serum
antibody product type
primary antibodies
clone
polyclonal
species reactivity
human
manufacturer/tradename
Chemicon®
technique(s)
ELISA: suitable
immunohistochemistry: suitable
NCBI accession no.
UniProt accession no.
shipped in
dry ice
target post-translational modification
unmodified
Gene Information
human ... GALR1(2587)
Specificity
Recognizes human Galanin. It is expected that the antibody will cross react with rat and mouse and other species due to the fact that galanin is extremely conserved.
Immunogen
Synthetic peptide from human Galanin. The immunogen peptide is available for use in absorbtion studies (Catalog Number AG370).
Application
Research Category
Neuroscience
Neuroscience
Research Sub Category
Neurotransmitters & Receptors
CNS Control of Metabolism
Neurotransmitters & Receptors
CNS Control of Metabolism
This Anti-Galanin Antibody is validated for use in ELISA, IH for the detection of Galanin.
Western blot: 1:1000-1:5000 using ECL
Immunohistochemistry
ELISA: 1:10,000-1:100,000 using 50-100 ng/well control peptide (Catalog Number AG370) per well.
Optimal working dilutions must be determined by the end user.
Immunohistochemistry
ELISA: 1:10,000-1:100,000 using 50-100 ng/well control peptide (Catalog Number AG370) per well.
Optimal working dilutions must be determined by the end user.
Target description
The unprocessed precursor is 13.3 kDa, the mature form is 3.16 kDa
Linkage
Replaces: AB1985
Physical form
Rabbit serum. Liquid with 0.05% sodium azide.
Storage and Stability
Maintain at -20°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles.
Legal Information
CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Scientific reports, 9(1), 5025-5025 (2019-03-25)
Spexin (SPX) is a highly conserved neuropeptide that is widely expressed in mammalian brain and peripheral tissue. In teleost, SPX1 is mainly expressed in the brain and ovary, and is involved in reproduction and food intake. A second form of
Current biology : CB, 32(7), 1497-1510 (2022-02-28)
Animals possess neuronal circuits inducing stress to avoid or cope with threats present in their surroundings, for instance, by promoting behaviors, such as avoidance and escape. However, mechanisms must exist to tightly control responses to stressors, since overactivation of stress
International journal of molecular sciences, 19(9) (2018-08-30)
The ileocecal valve (ICV)-a sphincter muscle between small and large intestine-plays important roles in the physiology of the gastrointestinal (GI) tract, but many aspects connected with the innervation of the ICV remain unknown. Thus, the aim of this study was
Journal of neurochemistry, 140(6), 889-902 (2016-07-29)
Kalirin (Kal), a dual Rho GDP/GTP exchange factor (GEF), plays essential roles within and outside the nervous system. Tissue-specific, developmentally regulated alternative splicing generates isoforms with one (Kal7) or two (Kal9, Kal12) GEF domains along with a kinase (Kal12) domain;
International journal of molecular sciences, 22(24) (2021-12-25)
Although guanethidine (GUA) was used in the past as a drug to suppress hyperactivity of the sympathetic nerve fibers, there are no available data concerning the possible action of this substance on the sensory component of the peripheral nervous system
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service