Skip to Content
Merck
All Photos(2)

Key Documents

AB2233

Sigma-Aldrich

Anti-Galanin Antibody

serum, from rabbit

Synonym(s):

galanin prepropeptide, galanin-message-associated peptide, galanin-related peptide

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

rat

species reactivity (predicted by homology)

mouse, boar, human

technique(s)

immunohistochemistry: suitable (paraffin)

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... GAL(51083)

General description

Galanin is a neuropeptide which is not a member of any known family of neuropeptides, despite repeated efforts to discover related peptides. Its actions are mediated via Gi-protein-coupled receptors (GPCR’s) and ion channels, usually producing inhibition of secretion of a transmitter or hormone in the nervous and endocrine system. In many respects, these inhibitory actions of galanin remind us of those of gamma-aminobutyric acid (GABA) and of neuropeptide Y (NPY). Galanin coexists with GABA, noradrenaline, 5-hydroxytryptamine (5-HT), and NPY in several regions of the brain. Galanin and it’s receptor are over expressed in limbic region of the brain in patients suffering from Alzheimer′s disease. However it is not clear whether galanin exacerbates or is a protective response to the disease.

Specificity

Cat. # AB2233 recognizes Galanin
Reactivity with other species has not been determined.

Immunogen

Synthetic Linear Peptide

Application

Detect Galanin using this Anti-Galanin Antibody validated for use in IH(P).
Research Category
Neuroscience
Research Sub Category
Neurochemistry & Neurotrophins

Quality

Routinely evaluated by Immunhistochemistry on rat cerebellum tissue.

Target description

13.3 kDa

Physical form

Serum with 0.05% NaN3

Storage and Stability

Maintain at -20°C for up to 1 year from date of receipt.

Analysis Note

Control
Rat Cerebellum Tissue

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yutaro Komuro et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 41(42), 8725-8741 (2021-09-01)
Galanin, one of the most inducible neuropeptides, is widely present in developing brains, and its expression is altered by pathologic events (e.g., epilepsy, ischemia, and axotomy). The roles of galanin in brain development under both normal and pathologic conditions have
Kamila Szymanska et al.
International journal of molecular sciences, 19(3) (2018-03-21)
Bisphenol A, used in the production of plastic, is able to leach from containers into food and cause multidirectional adverse effects in living organisms, including neurodegeneration and metabolic disorders. Knowledge of the impact of BPA on enteric neurons is practically
Liliana Rytel et al.
Anatomia, histologia, embryologia, 47(6), 517-526 (2018-08-15)
The gastrointestinal (GI) tract is innervated by nerve processes derived from the intramural enteric neurons and neurons localized outside the digestive tract. This study analysed the neurochemical characterization of nerves in the wall of the porcine oesophagus using single immunofluorescence
Krystyna Makowska et al.
Animals : an open access journal from MDPI, 10(12) (2021-01-10)
Bisphenol A (BPA) is widely utilized in plastic production process all over the world. Previous studies have shown that BPA, with its similarity to estrogen, may negatively affect living organisms. It is acknowledged that BPA distorts the activity of multiple
Katarzyna Palus et al.
Journal of chemical neuroanatomy, 72, 25-33 (2016-01-06)
The purpose of the present study was to determine the response of the porcine coeliac-superior mesenteric ganglion complex (CSMG) neurons projecting to the prepyloric area of the porcine stomach to peripheral neuronal damage following partial stomach resection. To identify the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service