Skip to Content
Merck
All Photos(2)

Key Documents

AB2251-I

Sigma-Aldrich

Anti-VGluT2 Antibody

serum, from guinea pig

Synonym(s):

Vesicular glutamate transporter 2, Differentiation-associated BNPI, Differentiation-associated Na(+)-dependent inorganic phosphate cotransporte, Solute carrier family 17 member 6, VGluT2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

guinea pig

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

rat, mouse

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

General description

Vesicular glutamate transporter 2 (UniProt Q9JI12; also known as Differentiation-associated BNPI, Differentiation-associated Na(+)-dependent inorganic phosphate cotransporte, Solute carrier family 17 member 6, VGluT2) is encoded by the Slc17a6 (also known as Dnpi, Vglut2) gene (Gene ID 84487) in rat species. Vesicular glutamate transporters (VGluTs) are proton-dependent carriers responsible for the upload of the major excitatory neurotransmitter glutamate into synaptic vesicles within presynaptic terminals to allow later regulated release at the synaptic cleft. Studies conducted in mice show that VGluT1 and VGluT2 are essential for vital functions, while Vglut3-knockout mice have a normal life expectancy despite being deaf and developing anxiety and discrete basal locomotor phenotypes. VgluT1- and VGluT2-expressing neurons represent the bulk of canonical glutamatergic neurons in the brain, with VgluT1 representing the major subtype and accounting for approximately 80% of total vesicular transport of glutamate in the brain. In contrast, VgluT3 is found in discrete population of non-glutamatergic neurons using other transmitters than glutamate, such as cholinergic interneurons in the dorsal and ventral striatum, subpopulations of GABAergic basket cells in the cortex and hippocampus, and serotoninergic neurons, where it is involved in more subtle modulation of local transmission. Rat VGluT2 is a 12-transmembrane (a.a. 72-92, 126-146, 149-169, 178-198, 217-237, 245-265, 311-331, 350-370, 387-407, 410-430, 444-464, 478-498) protein with 7 cytoplasmic and 6 vacuolar domains, having both its N- and C-terminal tails (a.a. 1-71 and 499-582) exposed at the cytoplasmic side.

Specificity

This polyclonal antibody targets VGluT2 C-terminal end sequence.

Immunogen

Epitope: C-terminus.
KLH-conjugated linear peptide corresponding to the C-terminal sequence of rat VGluT2.

Application

Immunohistochemistry Analysis: A 1:1,000 dilution from a representative lot detected VGluT2 in mouse and rat brain tissue sections.
Research Category
Neuroscience
Research Sub Category
Synapse & Synaptic Biology
This Anti-VGluT2 Antibody is validated for use in Western Blotting, Immunohistochemistry for the detection of VGluT2.

Quality

Evaluated by Western Blotting in rat brain membrane extract.

Western Blotting Analysis: A 1:10,000 dilution of this antibody detected VGluT2 in 10 µg of rat brain membrane extract.

Target description

~56 kDa observed. 64.58/64.56 kDa (rat/mouse) calculated. The smaller than predicted band size is consistent with the reported apparent molecular weight (Zhang, S., et al. (2015). Nat. Neurosci. 18(3):386-392; Moechars, D., et al. (2006). J. Neurosci. 26(46):12055-12066). Uncharacterized band(s) may appear in some lysates.

Physical form

Guinea pig polyclonal antibody serum with 0.05% sodium azide.
Unpurified.

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Natália Madeira et al.
Cerebral cortex (New York, N.Y. : 1991), 30(7), 4064-4075 (2020-03-13)
The acquisition of fear memories involves plasticity of the thalamic and cortical pathways to the lateral amygdala (LA). In turn, the maintenance of synaptic plasticity requires the interplay between input-specific synaptic tags and the allocation of plasticity-related proteins. Based on
Si-Qiang Ren et al.
Neuron, 104(2), 385-401 (2019-08-03)
The frontal area of the cerebral cortex provides long-range inputs to sensory areas to modulate neuronal activity and information processing. These long-range circuits are crucial for accurate sensory perception and complex behavioral control; however, little is known about their precise
Laura Modol et al.
Neuron, 105(1), 93-105 (2019-11-30)
The developmental journey of cortical interneurons encounters several activity-dependent milestones. During the early postnatal period in developing mice, GABAergic neurons are transient preferential recipients of thalamic inputs and undergo activity-dependent migration arrest, wiring, and programmed cell-death. Despite their importance for
Naoya Murao et al.
PloS one, 12(11), e0187213-e0187213 (2017-11-02)
Incretins (GLP-1 and GIP) potentiate insulin secretion through cAMP signaling in pancreatic β-cells in a glucose-dependent manner. We recently proposed a mechanistic model of incretin-induced insulin secretion (IIIS) that requires two critical processes: 1) generation of cytosolic glutamate through the
Damien Jullié et al.
Neuron, 105(4), 663-677 (2019-12-16)
A major function of GPCRs is to inhibit presynaptic neurotransmitter release, requiring ligand-activated receptors to couple locally to effectors at terminals. The current understanding of how this is achieved is through receptor immobilization on the terminal surface. Here, we show

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service