Skip to Content
Merck
All Photos(1)

Key Documents

AB15552

Sigma-Aldrich

Anti-AT1 Receptor Antibody

Chemicon®, from rabbit

Synonym(s):

Angiotensin II receptory type 1, AGTR1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse, rat, human

manufacturer/tradename

Chemicon®

technique(s)

flow cytometry: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... AGTR1(185)

Related Categories

Specificity

Recognizes AT1 Receptor (Angiotensin II receptory type 1, AGTR1).
The immunogen sequence is identical in canine.

Immunogen

Purified peptide from the N-terminus of human AT1 Receptor (Accession number P30556).

Application

Anti-AT1 Receptor Antibody detects level of AT1 Receptor & has been published & validated for use in FC, IC, IH & WB.
Research Category
Neuroscience
Research Sub Category
CNS Control of Metabolism

Hormones & Receptors
Western blot: 1:200-1:500 using ECL on rat liver, rat kidney, mouse heart and mouse kidney lysate.
Immunohistochemistry on rat brain sections.
Immunocytochemistry: 1:100
Flow cytometry, indirect: 5-10 µg of antibody /1 x 106 cells
Dilutions should be made using a carrier protein such as BSA (1-3%).
Optimal working dilutions must be determined by the end user.

Physical form

Format: Purified
ImmunoAffinity Purified
Lyophilized from phosphate buffered saline, pH 7.4, containing 1% BSA, and 0.05% sodium azide as a preservative. Reconstitute with 50 μL of sterile deionized water. Centrifuge antibody preparation before use (10,000 xg for 5 min).

Storage and Stability

Maintain lyophilized material at -20°C for up to 12 months after date of receipt. After reconstitution maintain at -20°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles.

Analysis Note

Control
Included free of charge with the antibody is 40 µg of control antigen (lyophilized powder). The stock solution of the antigen can be made up using 100 µL of sterile deionized water. For negative control, preincubate 1 µg of peptide with 1 µg of antibody for one hour at room temperature. Optimal concentrations must be determined by the end user.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Francielle Graus-Nunes et al.
World journal of hepatology, 11(4), 359-369 (2019-05-23)
Obesity has been associated with hepatic overexpression of the renin-angiotensin system (RAS). To evaluate the action of two angiotensin II (ANGII) receptor blockers (losartan or telmisartan) on the modulation of local hepatic RAS and the resulting metabolic effects in a
Mehmet Hakan Ozdener et al.
Nutrients, 14(13) (2022-07-10)
In addition to the α, β, and γ subunits of ENaC, human salt-sensing taste receptor cells (TRCs) also express the δ-subunit. At present, it is not clear if the expression and function of the ENaC δ-subunit in human salt-sensing TRCs
Julius Benicky et al.
Cellular and molecular neurobiology, 32(8), 1353-1365 (2012-07-31)
Commercially available Angiotensin II AT1 receptor antibodies are widely employed for receptor localization and quantification, but they have not been adequately validated. In this study, six commercially available AT1 receptor antibodies were characterized by established criteria: sc-1173 and sc-579 from
Hana Sutovska et al.
Experimental physiology, 106(8), 1762-1771 (2021-06-06)
What is the central question of this study? Artificial light at night decreases blood pressure and heart rate in rats. Are these changes in heart rate accompanied by changes in protein expression in the heart's left ventricle? What is the
K Semba et al.
Cell death & disease, 5, e1333-e1333 (2014-07-18)
Glaucoma, one of the leading causes of irreversible blindness, is characterized by progressive degeneration of optic nerves and retinal ganglion cells (RGCs). In the mammalian retina, excitatory amino acid carrier 1 (EAAC1) is expressed in neural cells, including RGCs, and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service