Skip to Content
Merck
All Photos(1)

Key Documents

A104-M

Millipore

NovaSeptum® Sealing

NovaSeptum® Manual Crimping Tool Spare Part Kit

Synonym(s):

Replacement Die for Novaseptum® Manual Crimping Tool

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104105
eCl@ss:
36101790
Pricing and availability is not currently available.

Quality Level

sterility

non-sterile

product line

NovaSeptum®

application(s)

sterile sampling

Looking for similar products? Visit Product Comparison Guide

Components

1 tube with pinch pipe for testing, 1 upper cutting tool

Legal Information

NOVASEPTUM is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service