Key Documents
Z114243
Supelco
Hand-operated aluminum cap crimper
O.D. 20 mm
Synonym(s):
Manual Cap Crimping Tool
About This Item
UNSPSC Code:
41122001
NACRES:
NB.11
O.D.
20 mm
General description
  • Used to attach aluminum seals to bottles and vials with a crimp/serum finish
  • Cushioned ergonomic handle reduces hand fatigue
  • Labeled for quick size identification
  • Polished crimping jaws provide consistent sealing
  • 2 year warranty
Choose from one of the most recent versions:
Certificates of Analysis (COA)