Skip to Content
Merck
All Photos(1)

Key Documents

8.43877

Sigma-Aldrich

Sodium tetrafluoroborate

for synthesis

Synonym(s):

Sodium tetrafluoroborate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
BF4Na
CAS Number:
Molecular Weight:
109.79
MDL number:
UNSPSC Code:
12352302
EC Index Number:
237-340-6
NACRES:
NA.22
Assay:
≥97.0% (acidimetric)
form:
crystals
Pricing and availability is not currently available.

Quality Level

Assay

≥97.0% (acidimetric)

form

crystals

mp

384 °C

storage temp.

2-30°C

SMILES string

[Na][B](F)(F)(F)F

InChI

1S/BF4.Na/c2-1(3,4)5;/q-1;+1

InChI key

KGJZTOFHXCFQIV-UHFFFAOYSA-N

Analysis Note

Assay (acidimetric): ≥ 97.0 %

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8B - Non-combustible, corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service