Skip to Content
Merck
All Photos(1)

Key Documents

8.43874

Sigma-Aldrich

1-Bromo-3-fluoro-4-(trifluoromethoxy)benzene

for synthesis

Synonym(s):

1-Bromo-3-fluoro-4-(trifluoromethoxy)benzene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H3BrF4O
CAS Number:
Molecular Weight:
259.00
MDL number:
UNSPSC Code:
12352005
EC Index Number:
430-850-1
NACRES:
NA.22

vapor pressure

4 hPa ( 25 °C)

Quality Level

Assay

≥99.0% (GC)

form

liquid

autoignition temp.

630 °C

potency

>2000 mg/kg LD50, oral (Rat)

bp

171 °C/994.5 hPa

mp

-55.5 °C

transition temp

flash point 69 °C

density

1.63 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

FC(F)(F)Oc1c(cc(cc1)Br)F

InChI

1S/C7H3BrF4O/c8-4-1-2-6(5(9)3-4)13-7(10,11)12/h1-3H

InChI key

SBSFDYRKNUCGBZ-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.708 - 1.712
Identity (IR): passes test

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Aquatic Acute 1 - Aquatic Chronic 1 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

156.2 °F

Flash Point(C)

69 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service