Skip to Content
Merck
All Photos(1)

Key Documents

8.40018

Sigma-Aldrich

N,N-Dimethylethylamine

for synthesis

Synonym(s):

N,N-Dimethylethylamine, Ethyldimethylamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H11N
CAS Number:
Molecular Weight:
73.14
MDL number:
UNSPSC Code:
12352116
EC Index Number:
209-940-8
NACRES:
NA.22
Pricing and availability is not currently available.

vapor pressure

527 hPa ( 20 °C)

Quality Level

Assay

≥99.0% (GC)

form

liquid

autoignition temp.

190 °C

potency

606 mg/kg LD50, oral (Rat)
>2000 mg/kg LD50, skin (Rabbit)

expl. lim.

0.9-11.2 % (v/v)

pH

12.0 (20 °C, 50 g/L in H2O)

bp

36.5 °C/1013 hPa

mp

-140 °C

transition temp

flash point -45.5 °C

density

0.675 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

N(CC)(C)C

InChI

1S/C4H11N/c1-4-5(2)3/h4H2,1-3H3

InChI key

DAZXVJBJRMWXJP-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.673 - 0.676
Identity (IR): passes test

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

-18.4 °F - closed cup

Flash Point(C)

-28 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service