Skip to Content
Merck
All Photos(2)

Key Documents

308102

Sigma-Aldrich

N,N-Dimethylhexylamine

98%

Synonym(s):

1-(Dimethylamino)hexane, N,N-Dimethyl-1-hexanamine, N,N-Dimethyl-n-hexylamine, N,N-Dimethylhexylamine, N-Hexyldimethylamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)4CH2N(CH3)2
CAS Number:
Molecular Weight:
129.24
Beilstein:
1732757
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

refractive index

n20/D 1.414 (lit.)

bp

146-150 °C (lit.)

density

0.744 g/mL at 25 °C (lit.)

functional group

amine

SMILES string

CCCCCCN(C)C

InChI

1S/C8H19N/c1-4-5-6-7-8-9(2)3/h4-8H2,1-3H3

InChI key

QMHNQZGXPNCMCO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The free Salinomycin (Sali), that has selective toxicity to cancer stem cells (CSCs), was encapsulated into the nanoparticle along with the additive N,N-dimethylhexylamine.

Application

N,N-Dimethylhexylamine was used as ion-pairing agent to study the problem of poor retention of uridine diphosphate-linked intermediates on reverse phase media. It was also used in the determining the five monophosphate nucleotides (cytidine 5′-monophosphate, uridine 5′-monophosphate, adenosine 5′-monophosphate, inosine 5′-monophosphate and guanosine 5′-monophosphate) in baby foods.

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

93.2 °F - closed cup

Flash Point(C)

34 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Peng Zhao et al.
Molecular pharmaceutics, 11(8), 2703-2712 (2014-06-25)
Salinomycin (Sali) has selective toxicity to cancer stem cells (CSCs), a subpopulation of cancer cells that have been recently linked with tumor multidrug resistance (MDR). To utilize its selective toxicity for cancer therapy, we sought to devise a nanoparticle (NP)
Pilar Viñas et al.
Journal of chromatography. A, 1217(32), 5197-5203 (2010-06-29)
A liquid chromatography with diode array detection coupled to dual electrospray atmospheric pressure chemical ionization time-of-flight mass spectrometry (HPLC/ESI-APCI-TOF-MS) method is described for the rapid determination of five monophosphate nucleotides (cytidine 5'-monophosphate, uridine 5'-monophosphate, adenosine 5'-monophosphate, inosine 5'-monophosphate and guanosine
Harika Vemula et al.
Analytical biochemistry, 465, 12-19 (2014-08-03)
Bacterial cell wall biosynthesis is the target of several antibiotics and is of interest as a target for new inhibitor development. The cytoplasmic steps of this pathway involve a series of uridine diphosphate (UDP)-linked peptidoglycan intermediates. Quantification of these intermediates

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service