Skip to Content
Merck
All Photos(1)

Key Documents

8.18606

Sigma-Aldrich

Triisobutyl phosphate

for synthesis

Synonym(s):

Triisobutyl phosphate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
((CH3)2CHCH2)3PO
CAS Number:
Molecular Weight:
266.31
MDL number:
UNSPSC Code:
12352108
EC Index Number:
204-798-3
NACRES:
NA.22

vapor pressure

0.002 hPa ( 20 °C)

Quality Level

Assay

≥99.0% (GC)

form

liquid

autoignition temp.

430 °C (DIN 51794)

potency

>5000 mg/kg LD50, oral (Rat)
>5000 mg/kg LD50, skin (Rabbit)

bp

272 °C/1013 hPa (decomposes)

mp

<-60 °C

transition temp

flash point 133 °C (DIN 51758)

solubility

0.26 g/L

density

0.965 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

[P](=O)(OCC(C)C)(OCC(C)C)OCC(C)C

InChI

1S/C12H27O4P/c1-10(2)7-14-17(13,15-8-11(3)4)16-9-12(5)6/h10-12H,7-9H2,1-6H3

InChI key

HRKAMJBPFPHCSD-UHFFFAOYSA-N

Related Categories

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.963 - 0.967
Identity (IR): passes test

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

270.5 °F - closed cup

Flash Point(C)

132.5 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service