71753-M
LB Broth, Miller - Novagen
Novagen®, granular, pack of 500 g or 5 kg, for Escherichia coli
Synonym(s):
Luria-Bertani broth, Lysogeny broth
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41106213
NACRES:
NA.54
Recommended Products
form
granular
Quality Level
manufacturer/tradename
Novagen®
packaging
pack of 500 g or 5 kg
storage condition
OK to freeze
technique(s)
microbiological culture: suitable
application(s)
microbiology
storage temp.
10-30°C
suitability
Escherichia coli
Related Categories
General description
LB Broth MILLER is a granulated medium for the cultivation of E. coli on scales ranging from small cultures to fermentation. The granules ensure rapid and uniform dissolution in water, and prevent clumping of the medium and inhalation of the airborne powder. It is supplied in several formats.
Application
LB Broth, Miller - Novagen has been used as a culture medium for Escherichia coli, Staphylococcus aureus, Pseudomonas aeruginosa strains.
Components
LB Broth, Miller contains yeast extract(5g), peptone from casein (10 g), and sodium chloride(10 g)
Warning
Toxicity: Standard Handling (A)
Preparation Note
EasyPak is an aluminum foil pouch containing 25 g granulated medium sufficient for 1 L culture. Just add the EasyPak contents to 1 L water, microwave for 2 min to sterilize*, and it is ready to use.
*Microwaving is sufficient if an antibiotic will be added to the prepared media.
*Microwaving is sufficient if an antibiotic will be added to the prepared media.
Legal Information
71753-5, 71753-6 and 71753-7 are not available for sale in Austria, Belgium, Denmark, Finland, France, Germany, Ireland, Italy, Luxembourg, Netherlands, Norway, Portugal, Spain, Sweden, Switzerland, and UK.
NOVAGEN is a registered trademark of Merck KGaA, Darmstadt, Germany
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 1
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Marcin Borowicz et al.
Journal of microbiological methods, 204, 106656-106656 (2022-12-17)
Crystal violet-based assay is widely used to evaluate the early stages of bacterial biofilm formation. Here, we modified it to study biofilm formation in medical tubing. While cost-effective and requiring no sophisticated equipment, our method provided quantitative and qualitative distinctions
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service