Skip to Content
Merck
All Photos(1)

Key Documents

539138

Millipore

Protease Inhibitor Cocktail VII

liquid, for the inhibition of serine proteases, cysteine proteases, aspartic proteases and thermolysin-like proteases and aminopeptidases. Recommended for purification of proteins containing His•Tag sequences.

Synonym(s):

Protease inhibitor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.54

product name

Protease Inhibitor Cocktail Set VII, Protease Inhibitor Cocktail Set VII contains five protease inhibitors with broad specificity. Recommended for purification of proteins containing His•Tag sequences.

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

shipped in

wet ice

storage temp.

−20°C

General description

A cocktail of five protease inhibitors with broad specificity for the inhibition of cysteine, serine, aspartic, and thermolysin-like proteases and aminopeptidases. This cocktail is recommended for purification of proteins containing His•Tag sequences. Each vial contains 100 mM AEBSF, HCl (Cat. No. 101500), 5 mM Bestatin (Cat. No. 200484), 1.5 mM E-64 (Cat. No. 324890), 2 mM Pepstatin A (Cat. No. 516481), and 200 µM of Phosphoramidon, Disodium Salt (Cat. No. 525276). One ml is recommended for the inhibition of proteases in 10 g of cells. Supplied with a data sheet. Note: 1 set = 5 x 1 ml.
This set is a specially formulated protease inhibitor cocktail of five protease inhibitors with broad specificity for the inhibition of cysteine, serine, aspartic, and thermolysin-like proteases and aminopeptidases. This cocktail is recommended for purification of histidine-tagged proteins.



This product is provided as a single vial or as a set of 5 vials. Each vial contains 1 ml of protease inhibitor cocktail solubilized in DMSO with the following components:

Specificity

Inhibits serine proteases, cysteine proteases, aspartic proteases and thermolysin-like proteases and aminopeptidases.

Warning

Toxicity: Irritant (B)

Physical form

In 1 ml DMSO.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

188.6 °F

Flash Point(C)

87 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tong Wang et al.
Methods in molecular biology (Clifton, N.J.), 2473, 101-128 (2022-07-13)
The entanglement of long axons found in cultured dissociated hippocampal neurons restricts the analysis of the machinery underlying directed axonal trafficking. Further, hippocampal neurons exhibit "en passant" presynapses that may confound the analysis of long-range retrograde axonal transport. To solve
Christopher R M Asquith et al.
Molecules (Basel, Switzerland), 23(5) (2018-05-23)
We demonstrate for the first time that 4H-1,2,6-thiadiazin-4-one (TDZ) can function as a chemotype for the design of ATP-competitive kinase inhibitors. Using insights from a co-crystal structure of a 3,5-bis(arylamino)-4H-1,2,6-thiadiazin-4-one bound to calcium/calmodulin-dependent protein kinase kinase 2 (CaMKK2), several analogues
Katrina M Piemonte et al.
The Journal of biological chemistry, 299(7), 104834-104834 (2023-05-19)
Chromatin organization is highly dynamic and modulates DNA replication, transcription, and chromosome segregation. Condensin is essential for chromosome assembly during mitosis and meiosis, as well as maintenance of chromosome structure during interphase. While it is well established that sustained condensin
Lindsey J Anstine et al.
Cancer research, 83(7), 997-1015 (2023-01-26)
Breast cancer subtypes and their phenotypes parallel different stages of the mammary epithelial cell developmental hierarchy. Discovering mechanisms that control lineage identity could provide novel avenues for mitigating disease progression. Here we report that the transcriptional corepressor TLE3 is a
Yan Wang et al.
Cell reports. Medicine, 5(6), 101580-101580 (2024-05-23)
Natural killer (NK) cell-based immunotherapy holds promise for cancer treatment; however, its efficacy remains limited, necessitating the development of alternative strategies. Here, we report that venetoclax, an FDA-approved BCL-2 inhibitor, directly activates NK cells, enhancing their cytotoxicity against acute myeloid

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service