Skip to Content
Merck
All Photos(1)

Key Documents

539133

Millipore

Protease Inhibitor Cocktail VI

liquid, for the inhibition of proteases and esterases. This small molecule/inhibitor is primarily used for Protease Inhibitors applications.

Synonym(s):

Protease inhibitor cocktail

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.54
form:
liquid
storage temp.:
−20°C

Product Name

Protease Inhibitor Cocktail Set VI, The Protease Inhibitor Cocktail Set VI controls the activity of Protease. This small molecule/inhibitor is primarily used for Protease Inhibitors applications.

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

shipped in

wet ice

storage temp.

−20°C

General description

A cocktail of six protease inhibitors with broad specificity for the inhibition of aspartic, cysteine, serine, and metalloproteases as well as aminopeptidases. This cocktail is recommended for use with plant cell extracts. One ml is recommended for 30 g of various plant tissues.



This cocktail is provided as a single vial of 1 ml, or as a package of 5 x 1 ml vials. Each vial contains the inhibitors solubilized in DMSO at the following concentrations:
The Protease Inhibitor Cocktail Set VI controls the activity of Protease. This small molecule/inhibitor is primarily used for Protease Inhibitors applications.

Specificity

Inhibits a broad spectrum of serine proteases, cysteine proteases, aspartic proteases, and metalloproteases, as well as aminopeptidases.

Biochem/physiol Actions

Primary Target
Aspartic, cysteine, serine, and metalloproteases as well as aminopeptidases.
Product does not compete with ATP.

Warning

Toxicity: Toxic (F)

Physical form

In 1 ml DMSO.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

188.6 °F

Flash Point(C)

87 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Huimeng Tang et al.
Food chemistry, 311, 126009-126009 (2019-12-31)
Proteomics and metabolomics were used to study the changes in proteins and metabolites in tomato fruits at different ripening stages and the effect of salt treatment on fruit quality. The results showed 2607 and 153 differentially expressed proteins in ripe
Weiqun Hu et al.
Bioscience reports, 40(5) (2020-05-28)
The study explored the effect of miR-30e-5p on nasopharyngeal carcinoma (NPC). MiR-30e-5p levels in NPC cancer and adjacent normal samples, in metastatic and non-metastatic cancer samples of NPC, and in NP69 cell and five NPC cell lines were determined by
Mousa Vatanmakanian et al.
Frontiers in endocrinology, 14, 1156120-1156120 (2023-10-30)
Prostate cancer (PCa) presents a significant health challenge in men, with a substantial number of deaths attributed to metastatic castration resistant PCa (mCRPC). Moreover, African American men experience disproportionately high mortality rates due to PCa. This study delves into the
Mingyue Li et al.
Cell death discovery, 9(1), 127-127 (2023-04-15)
Sestrins are a small gene family of pleiotropic factors whose actions promote cell adaptation to a range of stress conditions. In this report we disclose the selective role of Sestrin2 (SESN2) in dampening aerobic glycolysis to adapt to limiting glucose
Salim Abdisalaam et al.
Nucleic acids research, 50(5), 2681-2699 (2022-02-22)
Cyclic guanosine monophosphate-adenosine monophosphate synthase (cGAS) is activated in cells with defective DNA damage repair and signaling (DDR) factors, but a direct role for DDR factors in regulating cGAS activation in response to micronuclear DNA is still poorly understood. Here

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service