Skip to Content
Merck
All Photos(1)

Key Documents

401487

Sigma-Aldrich

IKK-2 Inhibitor VIII

The IKK-2 Inhibitor VIII, also referenced under CAS 406208-42-2, controls the biological activity of IKK-2. This small molecule/inhibitor is primarily used for Inflammation/Immunology applications.

Synonym(s):

IKK-2 Inhibitor VIII, 2-Amino-6-(2-(cyclopropylmethoxy)-6-hydroxyphenyl)-4-(4-piperidinyl)-3-pyridinecarbonitrile, ACHP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H24N4O2
CAS Number:
Molecular Weight:
364.44
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77
Pricing and availability is not currently available.

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

yellow

solubility

DMSO: 10 mg/mL

shipped in

wet ice

storage temp.

−20°C

SMILES string

N1CCC(CC1)c2c(c(nc(c2)c3c(cccc3O)OCC4CC4)N)C#N

InChI

1S/C21H24N4O2/c22-11-16-15(14-6-8-24-9-7-14)10-17(25-21(16)23)20-18(26)2-1-3-19(20)27-12-13-4-5-13/h1-3,10,13-14,24,26H,4-9,12H2,(H2,23,25)

InChI key

DYVFBWXIOCLHPP-UHFFFAOYSA-N

General description

A cell-permeable piperidinyl-pyridine compound that acts as a selective inhibitor of IKK-2 activity (IC50 = 8.5 and 250 nM for IKK-2 and IKK-1, respectively) with little effect towards IKK-3, Syk, and MKK4 (IC50 >20 µM). Exhibits good aqueous solubility (0.12 mg/ml in pH 7.4 isotonic buffer) and potent activity in various cellular assays in vitro (IC50<150 nM). Shown to be orally bioavailable both in rats and mice (BA = 60% and 16%, respectively) and effectively inhibit arachidonic acid-induced swelling in a murine ear edema model in vivo.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
IKK-2
Product does not compete with ATP.
Reversible: no
Target IC50: 8.5 and 250 nM for IKK-2 and IKK-1, respectively

Packaging

Packaged under inert gas

Warning

Toxicity: Irritant (B)

Preparation Note

Sonication is required for complete solubilization.

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Murata, T., et al. 2004. Bioorg. Med. Chem. Lett.14, 4019.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jin Hua Liang et al.
mBio, 12(4), e0153021-e0153021 (2021-07-21)
Epstein-Barr virus (EBV) is associated with 200,000 cancers annually, including B-cell lymphomas in immunosuppressed hosts. Hypomorphic mutations of the de novo pyrimidine synthesis pathway enzyme cytidine 5' triphosphate synthase 1 (CTPS1) suppress cell-mediated immunity, resulting in fulminant EBV infection and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service