Skip to Content
Merck
All Photos(6)

Key Documents

KITALYSIS-SM

Sigma-Aldrich

KitAlysis High-Throughput Suzuki-Miyaura Cross-Coupling Reaction Screening Kit - Pack of 4

Synonym(s):

Suzuki-Miyaura Cross-Coupling

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161600
NACRES:
NA.22

reaction suitability

reaction type: Cross Couplings
reaction type: Suzuki-Miyaura Coupling
reagent type: catalyst
core: palladium

functional group

phosphine

Application

Screening kit for Suzuki-Miyaura cross-coupling reaction. Step-by-step user guide and a downloadable experimental excel sheets are provided in the below hyperlink.
Step-by-Step Guide for Suzuki-Miyaura Cross-Coupling Reaction Screening Kit
Downloadable experimental excel sheet

Each kit contains 4 customizable screening sets that include the following components:
  • 6 x 4 pre-weighed catalysts in glass vials loaded with stir bars and topped with cap mat.
  • 1 (2 mL) ampule each of Toluene, THF, n-Butanol, DMAc, and aq. 1.5 M K3PO4.
  • 4 NEW KitAlysis 24-Well Reaction Block Replacement Films.
  • 4 substrate reaction vials with stir bars.
  • 1 internal standard.
Monitor reaction progress via TLC-MS by following this protocol

Best when used with KitAlysis Benchtop Inertion Box and 24-well Reaction Block (sold separately).

Features and Benefits

  • Designed and tested by synthetic chemists.
  • Easily run 24-microscale reactions in parallel.
  • 10 μmol reactions with 1 μmol catalyst, 3 equiv. base, 0.1 M concentration.
  • Customizable Excel Template for quick calculations and easy incorporation into electronic laboratory notebooks.

Legal Information

KitAlysis is a trademark of Sigma-Aldrich Co. LLC

Kit Components Only

Product No.
Description

  • SnapIT Ampule Opener, Regular, for 1-2ml, 5-10ml, 10-15ml ampoules 1 EA

Kit Components Also Available Separately

Product No.
Description
SDS

  • 732117Pd-PEPPSI-IPent catalyst, ≥95% 5 µmolSDS

  • 753009SPhos Pd G2 5 µmolSDS

  • 761311cataCXium® A Pd G2 5 µmolSDS

  • 763381XPhos Pd G3, 98%, 1:1 MTBE adduct 5 µmolSDS

  • 764175P(Cy3) Pd G3, 97% 5 µmolSDS

  • 764183APhos Pd G3, 97% 5 µmolSDS

  • 900522Toluene, anhydrous, ZerO2®, 99.8% 8 mLSDS

  • 900636Tetrahydrofuran, anhydrous, inhibitor-free, ZerO2®, ≥99.9% 8 mLSDS

  • 9006391-Butanol, anhydrous, ZerO2®, 99.8% 8 mLSDS

  • 900634N,N-Dimethylacetamide, anhydrous, ZerO2®, 99.8% 8 mLSDS

  • Degassed Potassium phosphate tribasic solution 8 mL

  • Z118850Spinbar® magnetic stirring fleas, red, L 10 mm, diam. 3 mm 1 EASDS

See All (12)

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Carc. 2 - Eye Dam. 1 - Flam. Liq. 2 - Repr. 1B - Skin Irrit. 2 - STOT RE 2 - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

Flash Point(F)

1.4 °F

Flash Point(C)

-17 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Scientist-Led High-Throughput Experimentation (HTE) and Its Utility in Academia and Industry.
Schmink JR, et al.
Aldrichimica Acta, 46, 71-80 (2013)
Spencer D Dreher et al.
Journal of the American Chemical Society, 130(29), 9257-9259 (2008-06-28)
Microscale parallel experimentation was used to discover three catalyst systems capable of coupling secondary organotrifluoroborates with sterically and electronically demanding aryl chlorides and bromides. The ensuing results represent the first comprehensive study of alkylboron coupling to aryl chlorides and, in
Karl D Collins et al.
Nature chemistry, 6(10), 859-871 (2014-09-23)
New organic reactivity has often been discovered by happenstance. Several recent research efforts have attempted to leverage this to discover new reactions. In this Review, we attempt to unify reported approaches to reaction discovery on the basis of the practical
High-Throughput Experimentation in Catalysis and Materials Science.
Gordillo A, et al.
Ullmann's Encyclopedia of Industrial Chemistry (2014)

Articles

All of the preformed catalysts used in the kit are air and moisture stable complexes in their commercially available form.

Tools aid in kit setup for organic chemistry techniques, ensuring ease and success.

Protocols

A step-by-step protocol guide for KitAlysis Suzuki-Miyaura Cross-Coupling Reaction Screening Kit.

A step-by-step protocol guide for KitAlysis Suzuki-Miyaura Cross-Coupling Reaction Screening Kit.

A step-by-step protocol guide for KitAlysis Suzuki-Miyaura Cross-Coupling Reaction Screening Kit.

A step-by-step protocol guide for KitAlysis Suzuki-Miyaura Cross-Coupling Reaction Screening Kit.

Related Content

Chemical reaction design and optimization in organic synthesis aims to precisely alter reaction parameters to achieve optimum results in finding pathways to target molecules.

Chemical reaction design and optimization in organic synthesis aims to precisely alter reaction parameters to achieve optimum results in finding pathways to target molecules.

Chemical reaction design and optimization in organic synthesis aims to precisely alter reaction parameters to achieve optimum results in finding pathways to target molecules.

Chemical reaction design and optimization in organic synthesis aims to precisely alter reaction parameters to achieve optimum results in finding pathways to target molecules.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service