Skip to Content
Merck
All Photos(1)

Key Documents

382113

Sigma-Aldrich

C646

≥99% (sum of isomers, HPLC), solid, histone acetyltransferase p300 inhibitor, Calbiochem®

Synonym(s):

Histone Acetyltransferase p300 Inhibitor, C646, 4-(4-{[5-(4,5-dimethyl-2-nitrophenyl)furan-2-yl]methylidene}-3-methyl-5-oxo-4,5-dihydro-1H-pyrazol-1-yl)benzoic acid, p300/CBP Inhibitor IV, Histone Acetyltransferase Inhibitor V, HAT Inhibitor V

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H19N3O6
CAS Number:
Molecular Weight:
445.42
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

product name

Histone Acetyltransferase p300 Inhibitor, C646, Histone Acetyltransferase p300 Inhibitor, C646, CAS 328968-36-1, is a cell-permeable, reversible inhibitor of p300/CBP HAT (Ki = 400 nM). Competes with acetyl-CoA for the p300 Lys-CoA binding pocket.

Quality Level

Assay

≥99% (sum of isomers, HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

brick red

solubility

DMSO: 50 mg/mL

shipped in

ambient

storage temp.

−20°C

InChI

1S/C24H19N3O6/c1-13-10-20(21(27(31)32)11-14(13)2)22-9-8-18(33-22)12-19-15(3)25-26(23(19)28)17-6-4-16(5-7-17)24(29)30/h4-12H,1-3H3,(H,29,30)/b19-12-

InChI key

HEKJYZZSCQBJGB-UNOMPAQXSA-N

General description

A reversible, cell-permeable pyrazolone p300/CBP HAT inhibitor (Ki = 400 nM), which competes with acetyl-CoA for the p300 Lys-CoA binding pocket. This compound demonstrates 86% inhibition against p300 at 10 nM, but elicits less than 10% inhibition against serotonin N-acetyltransferase, PCAF histone acetyltransferase, GCN5 histone acetyletransferase, Rtt109 histone acetyletransferase, Sas histone acetyl transferase, and MOZ histone acetyltransferase in a chemical screening assay. Treatment of C3H 10T1/2 mouse fibroblasts with 25 µM of compound results in an inhibitory effect against basal and TSA-inducible acetylation of histones H3 and H4. It is also shown to inhibit human cell growth in both melanoma and non-small-cell-lung (NSCL) cancer cell lines at 10 µM with similar or higher potency compared with that of the peptide-based bisubstrate p300/CBP HAT inhibitor Lys-CoA-Tat at 25 µM.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C. Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Bowers, EM., et al. 2010. Chem Biol17, 471.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daniel J Puleston et al.
Cell, 184(16), 4186-4202 (2021-07-04)
Polyamine synthesis represents one of the most profound metabolic changes during T cell activation, but the biological implications of this are scarcely known. Here, we show that polyamine metabolism is a fundamental process governing the ability of CD4+ helper T cells (TH)
Lu Yang et al.
Communications biology, 4(1), 1373-1373 (2021-12-10)
Tumor suppressor p53, a critical regulator of cell fate, is frequently mutated in cancer. Mutation of p53 abolishes its tumor-suppressing functions or endows oncogenic functions. We recently found that p53 binds via its proline-rich domain to peptidase D (PEPD) and
Virender Kumar Sharma et al.
Scientific reports, 11(1), 16427-16427 (2021-08-14)
Api5, is a known anti-apoptotic and nuclear protein that is responsible for inhibiting cell death in serum-starved conditions. The only known post-translational modification of Api5 is acetylation at lysine 251 (K251). K251 acetylation of Api5 is responsible for maintaining its
Kenji Ohshima et al.
The Journal of pathology, 256(2), 164-173 (2021-10-27)
Cancer cells have an altered metabolic state that supports their growth, for example, aerobic glycolysis, known as the Warburg effect. Colorectal cancer cells have been reported to exhibit the Warburg effect and mainly rely on glycolysis for progression and have
Kee-Beom Kim et al.
Science advances, 8(7), eabl4618-eabl4618 (2022-02-17)
EP300, a transcription coactivator important in proliferation and differentiation, is frequently mutated in diverse cancer types, including small cell lung cancer (SCLC). While these mutations are thought to result in loss of EP300 function, the impact on tumorigenesis remains largely

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service