Skip to Content
Merck
All Photos(1)

Key Documents

217691

Sigma-Aldrich

CDC25 Phosphatase Inhibitor I, BN82002

The CDC25 Phosphatase Inhibitor I, BN82002, also referenced under CAS 396073-89-5, controls the biological activity of CDC25 Phosphatase. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

CDC25 Phosphatase Inhibitor I, BN82002, N-(2-Hydroxy-3-methoxy-5-dimethylamino)benzyl, Nʹ-(2-(4-nitrophenethyl)), Nʹʹ-methylamine, PTP Inhibitor XX

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H25N3O4
CAS Number:
Molecular Weight:
359.42
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77
Pricing and availability is not currently available.

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

red to dark red

solubility

DMSO: 10 mg/mL
ethanol: 10 mg/mL

shipped in

ambient

storage temp.

−20°C

SMILES string

[N+](=O)([O-])c1ccc(cc1)CCN(Cc2c(c(cc(c2)N(C)C)OC)O)C

InChI

1S/C19H25N3O4/c1-20(2)17-11-15(19(23)18(12-17)26-4)13-21(3)10-9-14-5-7-16(8-6-14)22(24)25/h5-8,11-12,23H,9-10,13H2,1-4H3

InChI key

GOKYHQGRIIXMNE-UHFFFAOYSA-N

General description

A cell-permeable ortho-hydroxybenzylamino compound that displays anti-tumor properties. Acts as a potent, selective, and irreversible inhibitor of CDC25 phosphatase family (IC50 = 2.4, 3.9, 6.3, 5.4, and 4.6 µM for 25A, 25B2, 25B3, 25C, and 25C-cat, respectively). Displays ~20-fold greater selectivity for CDC25 phosphatases compared to CD45 tyrosine phosphatase. Shown to delay cell cycle progression in vitro (IC50 in the range of 7.2-32.6 µM), and reduces tumor growth in athymic mice xenografted with the human pancreatic cell MIA PaCa-2 (15 mg/kg, i.p. route).

Biochem/physiol Actions

Cell permeable: yes
Primary Target
CDC25A
Product does not compete with ATP.
Reversible: no
Target IC50: 2.4, 3.9, 6.3, 5.4, and 4.6 µM for CDC25 phosphatase family 25A, 25B2, 25B3, 25C, and 25C-cat, respectively

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C. Unstable in aqueous solution.

Other Notes

Brezak, M.C., et al. 2004. Cancer Res.64, 3320.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tom Kaufman et al.
Nature communications, 13(1), 2725-2725 (2022-05-19)
While multiplexing samples using DNA barcoding revolutionized the pace of biomedical discovery, multiplexing of live imaging-based applications has been limited by the number of fluorescent proteins that can be deconvoluted using common microscopy equipment. To address this limitation, we develop

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service