Skip to Content
Merck
All Photos(1)

Key Documents

20-180

Sigma-Aldrich

pNPP Tyr Assay Buffer

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116012
eCl@ss:
32160405
NACRES:
NA.41

form

liquid

Quality Level

manufacturer/tradename

Upstate®

technique(s)

activity assay: suitable (phosphatase)

shipped in

wet ice

Application

For use in the Tyrosine Assay Kit.

Physical form

25mM HEPES, pH 7.2, 50mM NaCl, 5mM dithiothreitol, 2.5mM EDTA

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Crystallization and preliminary X-ray diffraction studies of two domains of a bilobed extra-cytoplasmic function sigma factor SigC from Mycobacterium tuberculosis.
Krishan Gopal Thakur,B Gopal
Acta Crystallographica Section F, Structural Biology and Crystallization Communications null
J F Davis et al.
Avian diseases, 39(1), 162-174 (1995-01-01)
The clinical signs, hypoglycemia, and mortality of "spiking mortality syndrome" were experimentally reproduced. Seven groups of day-old male primary broiler breeder chicks were orally inoculated with tissue and/or fecal-urate homogenates taken from field broilers with spiking mortality syndrome and from
Thrombin and bradykinin initiate discrete endothelial solute permeability mechanisms.
R C Schaeffer,F Gong,M S Bitrick,T L Smith
The American Journal of Physiology null
J R Zalcberg et al.
International journal of cancer, 57(4), 522-528 (1994-05-15)
Separate mechanisms underlying the multidrug resistant (MDR) phenotype were identified in 2 independent approaches to select tumour cells resistant to low concentrations of doxorubicin (Dox) from the sensitive T cell leukemia cell line CCRF-CEM. The CEM/A7 cell line was selected

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service