Skip to Content
Merck
All Photos(1)

Key Documents

17-245

Sigma-Aldrich

Acetyl-Histone H3 Immunoprecipitation (ChIP) Assay Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
eCl@ss:
32161000
NACRES:
NA.84

manufacturer/tradename

Upstate®

Quality Level

technique(s)

immunoprecipitation (IP): suitable

NCBI accession no.

UniProt accession no.

Related Categories

General description

For use to immunoprecipitate transcriptionally active chromatin from mammalian cells using anti-acetyl-Histone H3. Detection of the gene or promoter of interest in immunoprecipitated chromatin must be empirically determined by the researcher using quantitative PCR or Southern slot-blot analysis with promotor specific primers or probe.

Application

For use to immunoprecipitate transcriptionally active chromatin from mammalian cells using anti-acetyl-Histone H3.

Packaging

Kit capacity: 22 assays

Components

Anti-acetyl-Histone H3 (Cat.# 06-599)
Protein A agarose/Salmon Sperm DNA (Cat.# 16-157)
All necessary buffers.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Irrit. 2

Storage Class Code

12 - Non Combustible Liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service