Skip to Content
Merck
All Photos(1)

Key Documents

1.04928

Supelco

Potassium carbonate

for analysis EMSURE® ACS,ISO,Reag. Ph Eur

Synonym(s):

Potassium carbonate, potash

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CK2O3
CAS Number:
Molecular Weight:
138.21
MDL number:
UNSPSC Code:
12352302
EC Index Number:
209-529-3
NACRES:
NA.21
Assay:
≥99.0% (acidimetric)
≥99.0% dry basis (acidimetric)
grade:
ACS reagent
form:
solid

grade

ACS reagent

Quality Level

Agency

reag. ISO
reag. Ph. Eur.

product line

EMSURE®

Assay

≥99.0% (acidimetric)
≥99.0% dry basis (acidimetric)

form

solid

potency

>2000 mg/kg LD50, oral (Rat)

impurities

≤0.005% Insoluble matter
≤0.01% Substances precipitated by ammonia

loss

≤1% loss on drying, 300 °C

pH

11.5-12.5 (20 °C, 50 g/L in H2O)

mp

891 °C

density

2.43 g/cm3 at 20 °C

bulk density

750 kg/m3

anion traces

chloride (Cl-): ≤0.003%
phosphate (PO43-): ≤0.001%
silicate (as SiO2): ≤0.005%
sulfur compounds (as SO42-): ≤0.003%

cation traces

Ca: ≤0.002%
Cu: ≤0.0005%
Fe: ≤0.0005%
Mg: ≤0.001%
N: ≤0.001%
Na: ≤0.02%
Pb: ≤0.0005%
heavy metals: ≤0.0005%

storage temp.

2-30°C

SMILES string

[K+].[K+].[O-]C(=O)[O-]

InChI

1S/CH2O3.2K/c2-1(3)4;;/h(H2,2,3,4);;/q;2*+1/p-2

InChI key

BWHMMNNQKKPAPP-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application


  • Catalytic pyrolysis and in situ carbonization of walnut shells: This research presents a method for the poly-generation of energy and enhanced electrochemical performance of carbons using potassium carbonate. The study highlights the application in biochar production and its potential use in energy storage and conversion devices, showcasing the integration of environmental sustainability and material science (He W et al., 2024).

  • Improved synthesis of 6-bromo-7-[(11)C]methylpurine for clinical use: This paper describes the enhanced synthesis process involving potassium carbonate, which is crucial for the preparation of radiopharmaceuticals used in medical imaging. This underscores the compound′s role in enhancing pharmaceutical research and diagnostics (Okamura T et al., 2024).

  • Effects of Xanthan gum and Potassium carbonate on the quality and flavor properties of frozen Jiuniang doughs: The study explores the use of potassium carbonate in food science, particularly in improving the texture and flavor of frozen dough products. It presents a potential application in food preservation and processing industries (Ma W et al., 2023).

  • Potential of GTL biosolids in a circular economy: This article investigates the use of potassium carbonate in the treatment and utilization of biosolids, highlighting its role in environmental sustainability and waste management. The research contributes to the development of greener technologies in the life science manufacturing sector (Zuhara S et al., 2023).

  • Potassium carbonate-mediated β-selective anomeric O-alkylation with primary electrophiles: The paper presents a method for synthesizing glycosphingolipids using potassium carbonate, emphasizing its importance in organic synthesis and pharmaceutical applications. This demonstrates the adaptability of potassium carbonate in facilitating complex chemical reactions within pharmaceutical research (Lakshika Hettiarachchi I et al., 2023).


Linkage

Replaces: PX1390; PX1390-1; PX1390-20

Analysis Note

Assay (acidimetric; calculated on dried substance): ≥ 99.0 %
Assay (acidimetric): ≥ 99.0 %
Insoluble matter: ≤ 0.005 %
Chloride (Cl): ≤ 0.003 %
Phosphate (PO₄): ≤ 0.001 %
Silicate (SiO₂): ≤ 0.005 %
Total sulfur (as sulfate): ≤ 0.003 %
Total nitrogen (N): ≤ 0.001 %
Heavy metals (ACS): ≤ 0.0005 %
Ca (Calcium): ≤ 0.002 %
Cu (Copper): ≤ 0.0005 %
Fe (Iron): ≤ 0.0005 %
Mg (Magnesium): ≤ 0.001 %
Na (Sodium): ≤ 0.02 %
Pb (Lead): ≤ 0.0005 %
Substances precipitated by ammonia: ≤ 0.01 %
Loss on drying (300 °C): ≤ 1.0 %
Corresponds to ACS, ISO, Reag. Ph Eur

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service