Skip to Content
Merck
All Photos(1)

Key Documents

1.01164

Supelco

Ammonium fluoride

for analysis EMSURE® ACS

Synonym(s):

Ammonium fluoride, Fluoroammonium

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
H4FN
CAS Number:
Molecular Weight:
37.04
MDL number:
UNSPSC Code:
12352301
EC Index Number:
235-185-9
NACRES:
NA.21
grade:
ACS reagent
form:
solid
solubility:
820 g/L

grade

ACS reagent

Quality Level

product line

EMSURE®

Assay

≥98.0% (NH4F basis, precipitation titration)

form

solid

impurities

≤0.005% Insoluble matter
≤0.5% Ammonium hydrogen difluoride
≤5% Water

ign. residue

≤0.01%

pH

6 (20 °C, 50 g/L in H2O)

solubility

820 g/L

density

1.01 g/cm3 at 20 °C

bulk density

250‑350 kg/m3

anion traces

chloride (Cl-): ≤0.001%
hexafluorosilicate (SiF62-): ≤0.1%
sulfate (SO42-): ≤0.005%

cation traces

Fe: ≤0.0005%
K: ≤0.002%
Na: ≤0.002%
heavy metals (as Pb): ≤0.0005%

storage temp.

15-25°C

SMILES string

F.F.N

InChI

1S/2FH.H3N/h2*1H;1H3

InChI key

KVBCYCWRDBDGBG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Ammonium Fluoride in Analytical Chemistry: Ammonium fluoride is extensively used in chromatographic analyses as demonstrated in the synthesis of bifunctional magnetic polymers for the pretreatment and determination of specific metabolites in plasma samples, enhancing both the selectivity and sensitivity of the detection processes (Sun et al., 2024).

  • Silica Synthesis: The compound is used in the activation mechanism for the synthesis of hydrated silica from industrial waste, showcasing its role in promoting environmental sustainability and material recovery (Duan et al., 2024).

  • Energy Storage Materials: In the field of energy storage, ammonium fluoride acts as a structuring agent in the systematic design of single metal compounds, which are crucial for developing advanced battery technologies (Kuo et al., 2023).

  • Environmental Safety and Photocatalysis: It also finds application in environmental safety and photocatalytic processes, where it is used to enhance the photocatalytic activity of materials for biomass conversion and CO2 reduction, contributing to cleaner energy solutions (Liu et al., 2023).

Linkage

Replaces: AX1295; AX1295-1

Analysis Note

Assay (precipitation titration, NH₄F): ≥ 98.0 %
Ammonium hydrogen difluoride (NH₄HF₂): ≤ 0.5 %
Chloride (Cl): ≤ 0.001 %
Hexafluorosilicate (SiF₆): ≤ 0.1 %
Sulfate (SO₄): ≤ 0.005 %
Heavy metals (as Pb): ≤ 0.0005 %
Fe (Iron): ≤ 0.0005 %
K (Potassium): ≤ 0.002 %
Na (Sodium): ≤ 0.002 %
Water: ≤ 5 %
Insoluble matter: ≤ 0.005 %
Residue on ignition: ≤ 0.01 %
Conform to ACS

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 1

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service