Skip to Content
Merck
All Photos(2)

Key Documents

338869

Sigma-Aldrich

Ammonium fluoride

≥99.99% trace metals basis

Synonym(s):

Ammonia Hydrofluoride, Azanium fluoride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH4F
CAS Number:
Molecular Weight:
37.04
EC Number:
MDL number:
UNSPSC Code:
12352302
eCl@ss:
38050106
PubChem Substance ID:
NACRES:
NA.23
grade:
for analytical purposes
form:
crystals and lumps

grade

for analytical purposes

Assay

≥99.99% trace metals basis

form

crystals and lumps

impurities

≤100 ppm METAL TRACE ANALYSIS (ICP)

SMILES string

N.F

InChI

1S/FH.H3N/h1H;1H3

InChI key

LDDQLRUQCUTJBB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ammonium fluoride is a white crystalline solid soluble in water. It adopts the wurtzite crystal structure and forms mixed crystals similar to ice. It is also known for its use as an energetic fluorinating agent and finds application in the field of solar cells and batteries.

Application

Ammonium fluoride can be used:
  • To modify the interface between the electron transport layer and the perovskite layer of solar cells. This helps to reduce interfacial defects and charge recombination enhancing power conversion efficiency of perovskite solar cells (PSCs).
  • As a structure directing agent to prepare electroactive materials for supercapacitors.
  • To synthesize NiF2/porous carbon (NPC) nanocomposite by a bottom-up method for cathodes of Li-ion battery. Heat treatment with NH4F produces a high yield of metal fluoride/porous carbon nanocomposites.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

does not flash

Flash Point(C)

does not flash

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bright, water-soluble CeF3 photo-, cathodo-, and X-ray luminescent nanoparticles.
Clement S, et al.
Journal of Nanoparticle Research, 17(1), 7-7 (2015)
Nanoparticle-mediated singlet oxygen generation from photosensitizers
Clement S, et al.
Journal of Photochemistry and Photobiology A: Chemistry, 332, 66-71 (2017)
Yukti Arora et al.
Scientific reports, 8(1), 12752-12752 (2018-08-26)
Storage of solar radiation is currently accomplished by coupling two separate devices, one that captures and converts the energy into an electrical impulse (a photovoltaic cell) and another that stores this electrical output (a battery or a supercapacitor electrochemical cell).
Xun, X. et al.
Chemistry of Materials, 9, 2966-2966 (1997)
Owen Kendall et al.
Nanomaterials (Basel, Switzerland), 10(5) (2020-05-06)
Fluorine-doped tin oxide (FTO) is one of the most studied and established materials for transparent electrode applications. However, the syntheses for FTO nanocrystals are currently very limited, especially for stable and well-dispersed colloids. Here, we present the synthesis and detailed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service