Skip to Content
Merck
All Photos(1)

Key Documents

05-554

Sigma-Aldrich

Anti-KDR/Flk-1/VEGFR2 Antibody, clone CH-11

clone CH-11, Upstate®, from mouse

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

CH-11, monoclonal

species reactivity

human, mouse

packaging

antibody small pack of 25 μg

manufacturer/tradename

Upstate®

technique(s)

ELISA: suitable
immunocytochemistry: suitable
western blot: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... KDR(3791)

Specificity

KDR/Flk-1

Immunogen

Recombinant protein corresponding to residues 1158-1345 of murine KDR/Flk-1

Application

Anti-KDR/Flk-1/VEGFR2 Antibody, clone CH-11 detects level of KDR/Flk-1/VEGFR2 & has been published & validated for use in ELISA, IC & WB.

Quality

routinely evaluated by immunoblot on RIPA lysates from HUVEC cells

Target description

230kDa

Physical form

20mM sodium phosphate, pH 7.6, 250mM NaCl with 0.1% sodium azide
Format: Purified

Other Notes

Please refer to lot specific datasheet.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ashok Aspatwar et al.
Journal of enzyme inhibition and medicinal chemistry, 35(1), 109-117 (2019-11-07)
With the aim to obtain novel compounds possessing both strong affinity against human carbonic anhydrases and low toxicity, we synthesised novel thiourea and sulphonamide derivatives 3, 4 and 10, and studied their in vitro inhibitory properties against human CA I, CA
Rebecca R Foster et al.
The American journal of pathology, 173(4), 938-948 (2008-09-06)
We have previously reported expression of vascular endothelial growth factor (VEGF)-A and -C in glomerular podocytes and actions of VEGF-A on glomerular endothelial cells (GEnC) that express VEGF receptor-2 (VEGFR-2). Here we define VEGFR-3 expression in GEnC and investigate the
Mária Báthori et al.
Journal of chromatographic science, 42(5), 275-279 (2004-06-11)
A suitable combination of preparative scale separation methods results in effective clean-up of the ecdysteroids of Silene italica ssp. nemoralis (Waldst. and Kit.) Nyman. The isolation of minor ecdysteroids from the partially purified extract is based on the use of
Nikola Burdzhiev et al.
Molecules (Basel, Switzerland), 25(17) (2020-08-23)
Tautomerization of 2-phenylimidazolecarbaldehydes has not been studied in detail so far, although this process is a well-known phenomenon for imidazole derivatives. That is why we focus our study on a series of 2-phenylimidazolecarbaldehydes and their parent alcohols that were synthesized
Aleksandar Pashev et al.
Beilstein journal of organic chemistry, 16, 1456-1464 (2020-07-11)
The Castagnoli-Cushman reaction of 3,4-dihydroisoquinolines with glutaric anhydride, its oxygen and sulfur analogues was investigated as a one-step approach to the benzo[a]quinolizidine system and its heterocyclic analogs. An extension towards the pyrrolo[2,1-a]isoquinoline system was achieved with the use of succinic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service