Skip to Content
Merck
All Photos(7)

Key Documents

07-352

Sigma-Aldrich

Anti-acetyl-Histone H3 (Lys9) Antibody

serum, Upstate®

Synonym(s):

H3K9Ac, Histone H3 (acetyl K9), H3 histone family, member T, histone 3, H3, histone cluster 3, H3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human, Saccharomyces cerevisiae, yeast, vertebrates

manufacturer/tradename

Upstate®

technique(s)

ChIP: suitable (ChIP-seq)
multiplexing: suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

acetylation (Lys9)

Gene Information

human ... H3C1(8350)

General description

Histone H3 is one of the five main histone proteins involved in the structure of chromatin in eukaryotic cells. Featuring a main globular domain and a long N-terminal tail, H3 is involved with the structure of the nucleosomes of the ′beads on a string′ structure. The N-terminal tail of histone H3 protrudes from the globular nucleosome core and can undergo several different types of epigenetic modifications that influence cellular processes. These modifications include the covalent attachment of methyl or acetyl groups to lysine and arginine amino acids and the phosphorylation of serine or threonine.

Specificity

Broad species-cross reactivity is expected.
Recognizes acetyl-histone H3 (Lys9), Mr 17 kDa. An additional unknown protein was detected at Mr 23 kDa.

Immunogen

Epitope: a.a. 4-14
Ovalbumin-conjugated, synthetic peptide (KQTARAcKSTGGK-C) corresponding to amino acids 4-14 of yeast histone H3 acetylated on lysine 9, with a C-terminal cysteine added for conjugation purposes.

Application

Anti-acetyl-Histone H3 (Lys9) Antibody is a Rabbit Polyclonal Antibody for detection of acetyl-Histone H3 (Lys9) also known as H3K9Ac, Histone H3 (acetyl K9) and has been published and validated in ChIP, WB, Mplex.
Chromatin Immunoprecipitation (ChIP):
An independent laboratory has shown this antibody preferentially immunoprecipitates chromatin from wild type yeast and not from a yeast strain containing a Lysine substitution to Alanine at residue 9.

Beadlyte Histone-Peptide Specificity Assay:
A 1:1000-1:5000 dilution of this lot was incubated with histone H3 peptides containing various modifications conjugated to Luminex microspheres. No cross-reactivity with peptides containing acetyl-lysine 14 or acetyl-lysine 27 was observed.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Histones

Quality

Routinely evaluated by western blot on acid extracts from sodium butyrate treated HeLa cells.

Western Blot Analysis:
A 1:5000-1:10,000 dilution of this lot detected acetyl-Histone H3 (Lys9) in acid extracts from sodium butyrate treated HeLa cells (Catalog # 17-305).

Target description

17 kDa

Linkage

Replaces: 04-1003

Physical form

Antiserum containing 0.35% sodium azide and 30% glycerol.
Unpurified

Storage and Stability

Stable for 1 year at -20°C from date of receipt. For maximum recovery of product, centrifuge the vial prior to removing the cap.

Analysis Note

Control
TSA-treated HeLa and NIH/3T3 cells, osteosarcoma tissue.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Genome-wide profiling of histone H3 lysine 9 acetylation and dimethylation in Arabidopsis reveals correlation between multiple histone marks and gene expression.
Zhou J, Wang X, He K, Charron JB, Elling AA, Deng XW
Plant Molecular Biology null
Genome-wide analysis of abnormal H3K9 acetylation in cloned mice.
Suzuki, T; Kondo, S; Wakayama, T; Cizdziel, PE; Hayashizaki, Y
Testing null
The transcription factor snail mediates epithelial to mesenchymal transitions by repression of estrogen receptor-alpha.
Dhasarathy, A; Kajita, M; Wade, PA
Molecular Endocrinology null
Transcription-dependent silencing of inducible convergent transgenes in transgenic mice.
Calero-Nieto, FJ; Bert, AG; Cockerill, PN
Epigenetics & Chromatin null
CBP/p300-mediated acetylation of histone H3 on lysine 56.
Das, C; Lucia, MS; Hansen, KC; Tyler, JK
Nature null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service