Skip to Content
Merck
All Photos(1)

Key Documents

04-1570-I

Sigma-Aldrich

Anti-RNA polymerase II subunit B1 (phospho-CTD Ser-7) Antibody, clone 4E12

culture supernatant, clone 4E12, from rat

Synonym(s):

DNA-directed RNA polymerase II subunit RPB1, RNA polymerase II subunit B1, DNA-directed RNA polymerase II subunit A, DNA-directed RNA polymerase III largest subunit, RNA-directed RNA polymerase II subunit RPB1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rat

Quality Level

antibody form

culture supernatant

antibody product type

primary antibodies

clone

4E12, monoclonal

species reactivity

mouse

species reactivity (predicted by homology)

human (based on 100% sequence homology)

technique(s)

ChIP: suitable
ELISA: suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

phosphorylation (pSer7)

Gene Information

human ... POLR2B(5431)

General description

RPB1 (RNA polymerase II subunit B1) is the catalytic component of RNA polymerase II which synthesizes mRNA and non-coding RNAs. During transcription, elongation is influenced by the phosphorylation status of the C-terminal domain (CTD) of RPB1, which acts as an assembly platform for factors regulating transcription initiation, elongation, termination and mRNA processing. Phosphorylation occurs mainly at residues ′Ser-2′ and ′Ser-5′ of the tandem 7 residue repeats in the C-terminal domain (CTD), which activates Pol II. This phosphorylation also can occur at ′Ser-7′ of the heptapepdtide repeat. This antibody recognizes the ′Ser-7′ CTD residue of RPB1.

Specificity

Demonstrated to react with human RBP1 in Ni et al., 2011. Transcription 2:5, 237-242
This antibody recognizes RNA polymerase II subunit B1 phosphorylated at Ser7 of the C-terminal domain (CTD).

Immunogen

Epitope: Phosphorylated Ser7 of the C-terminal domain (CTD)
Ovalbumin-conjugated linear peptide corresponding to human RNA polymerase II subunit B1 phosphorylated at Ser7 of the C-terminal domain (CTD).

Application

Anti-RNA polymerase II subunit B1 (phospho-CTD Ser-7) Antibody, clone 4E12 is a highly specific rat monoclonal antibody & that targets RNA Polymerase & has been tested in western blotting.
Chromatin Immunoprecipitation Analysis: A representative lot was used by an independent laboratory to immunoprecipitate RNA polymerase II subunit B1 (phospho-CTD Ser-5) in ChIP (Chapman, R., et al. (2007). Science. 318(5857):1780-1782.).
Demonstrated to react with human RBP1 in Ni et al., 2011. Transcription 2:5, 237-242

Quality

Evaluated by Western Blotting in untreated and lambda phosphatase treated NIH/3T3 cell lysate.

Western Blotting Analysis: A 1:2,000 dilution from a representative lot detected RNA polymerase II subunit B1 (phospho-CTD Ser-7) in 10 μg of untreated NIH/3T3 cell lysate.

Target description

~220 kDa observed

Linkage

Replaces: 04-1570

Physical form

Rat monoclonal IgG1κ with 0.05% sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Emily Brookes et al.
Cell stem cell, 10(2), 157-170 (2012-02-07)
Polycomb repressor complexes (PRCs) are important chromatin modifiers fundamentally implicated in pluripotency and cancer. Polycomb silencing in embryonic stem cells (ESCs) can be accompanied by active chromatin and primed RNA polymerase II (RNAPII), but the relationship between PRCs and RNAPII
Xiaohong Zhao et al.
Cell reports, 34(11), 108870-108870 (2021-03-18)
Ibrutinib, a bruton's tyrosine kinase (BTK) inhibitor, provokes robust clinical responses in aggressive mantle cell lymphoma (MCL), yet many patients relapse with lethal Ibrutinib-resistant (IR) disease. Here, using genomic, chemical proteomic, and drug screen profiling, we report that enhancer remodeling-mediated
Jing Gao et al.
Haematologica (2021-06-25)
Despite significant progress in the treatment of patients with diffuse large B-cell lymphoma (DLBCL) and mantle cell lymphoma (MCL), prognosis of patients with relapsed disease remains poor due to the emergence of drug resistance and subsequent disease progression. Identification of
Tanaz Sharifnia et al.
Nature medicine, 25(2), 292-300 (2019-01-22)
Chordoma is a primary bone cancer with no approved therapy1. The identification of therapeutic targets in this disease has been challenging due to the infrequent occurrence of clinically actionable somatic mutations in chordoma tumors2,3. Here we describe the discovery of
Baishan Jiang et al.
Nature chemical biology, 17(6), 675-683 (2021-03-24)
Cyclin-dependent kinase 12 (CDK12) is an emerging therapeutic target due to its role in regulating transcription of DNA-damage response (DDR) genes. However, development of selective small molecules targeting CDK12 has been challenging due to the high degree of homology between

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service