Skip to Content
Merck
All Photos(2)

Key Documents

F0287

Sigma-Aldrich

Fibronectin Proteolytic Fragment from human plasma

lyophilized powder, 70 kDa

Synonym(s):

Fibronectin

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202
eCl@ss:
42020141
NACRES:
NA.75

biological source

human plasma

Quality Level

Assay

>90% (SDS-PAGE)

form

lyophilized powder

mol wt

70 kDa

packaging

pkg of 0.5 mg

technique(s)

cell culture | mammalian: suitable

impurities

HIV and HBsAg, source material tested negative
Small proteolytic fragments, may contain traces

solubility

water: soluble ≥0.500 mg/mL, clear to slightly hazy, colorless

UniProt accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

human ... FN1(2335)

Looking for similar products? Visit Product Comparison Guide

General description

Fibronectin 1 is a glycoprotein of the extracellular matrix that is coded by FN1 gene. It is expressed in the plasma and at the cell surface. FN1 is mapped to human chromosome 2q35. Proteolytic fragments of fibronectin has a cell binding domain.

Application

Fibronectin Proteolytic Fragment from human plasma has been used in pre adsorbed antibody blocking assay and EC-pericyte tube coassembly.

Biochem/physiol Actions

Fibronectin participates in cell adhesion, growth, migration, wound healing, blood coagulation and metastasis. Mutations in FN1 results in glomerulopathy. It plays an important role in cell attachment and spreading, control of cell cytoskeleton, morphology and differentiation. FN1 is also involved in extracellular matrix formation, hemostasis and thrombosis. Proteolytic fragments of fibronectin plays a vital role in mononuclear phagocyte function.

Caution

The product should be stored at -20°C.

Preparation Note

This product is lyophilized from phosphate buffered saline with sucrose as a cryoprotectant. The source material has tested negative for antibody to HIV, HCN, and HBsAg. It is soluble in water at 0.5 mg/mL and yields a clear to slightly hazy solution.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mutations F352A and Y528A in human HSP90I? reduce fibronectin association and fibrillogenesis in cell-derived matrices.
Chakraborty, et al.
Cell Stress & Chaperones (2023)
Fibronectin in Health and Disease (2018)
C Penn et al.
Microbial pathogenesis, 17(6), 387-393 (1994-12-01)
Candida albicans yeast cells bind soluble human plasma fibronectin (Fn) through a glycoprotein receptor (adhesin) located on the cell surface. This work demonstrates that a 120 kDa proteolytic fragment of Fn encompassing the cell binding domain binds more avidly to
Pericyte recruitment during vasculogenic tube assembly stimulates endothelial basement membrane matrix formation.
Stratman A N, et al.
Blood, 114(24), 5091-5101 (2009)
Ail protein binds ninth type III fibronectin repeat (9FNIII) within central 120 kDa region of fibronectin to facilitate cell binding by Yersinia pestis.
Tsang T M, et al.
The Journal of Biological Chemistry, 287(20), 16759-16767 (2012)

Protocols

Dilute fibronectin for cell attachment, varying per cell type. Coating protocol, products, and FAQs provided.

Dilute fibronectin for cell attachment, varying per cell type. Coating protocol, products, and FAQs provided.

Dilute fibronectin for cell attachment, varying per cell type. Coating protocol, products, and FAQs provided.

Dilute fibronectin for cell attachment, varying per cell type. Coating protocol, products, and FAQs provided.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service