T-078
3,3′,5′-Triiodo-L-thyronine-13C6 (reverse T3-13C6)
100 μg/mL in methanol with 0.1N NH3, ampule of 1 mL, certified reference material, Cerilliant®
Synonym(s):
3,3′,5′-Triiodo-L-thyronine-13C6 solution
About This Item
Recommended Products
grade
certified reference material
Quality Level
form
liquid
feature
(Snap-N-Spike®)
packaging
ampule of 1 mL
manufacturer/tradename
Cerilliant®
concentration
100 μg/mL in methanol with 0.1N NH3
technique(s)
gas chromatography (GC): suitable
liquid chromatography (LC): suitable
format
single component solution
storage temp.
−20°C
SMILES string
IC1=CC(O[13C]2=[13CH][13CH]=[13C](C[C@H](N)C(O)=O)[13CH]=[13C]2I)=CC(I)=C1O
InChI
1S/C15H12I3NO4/c16-9-3-7(4-12(19)15(21)22)1-2-13(9)23-8-5-10(17)14(20)11(18)6-8/h1-3,5-6,12,20H,4,19H2,(H,21,22)/t12-/m0/s1/i1+1,2+1,3+1,7+1,9+1,13+1
InChI key
HZCBWYNLGPIQRK-CXGLLCBXSA-N
General description
Legal Information
related product
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1
Target Organs
Eyes,Central nervous system
Storage Class Code
3 - Flammable liquids
WGK
WGK 2
Flash Point(F)
49.5 °F
Flash Point(C)
9.7 °C
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Articles
Determination of free thyroid hormones (T3 & T4) in serum by LC-MS/MS following a streamlined 1-hour BioSPME sample preparation.
Determination of free thyroid hormones (T3 & T4) in serum by LC-MS/MS following a streamlined 1-hour BioSPME sample preparation.
Determination of free thyroid hormones (T3 & T4) in serum by LC-MS/MS following a streamlined 1-hour BioSPME sample preparation.
Determination of free thyroid hormones (T3 & T4) in serum by LC-MS/MS following a streamlined 1-hour BioSPME sample preparation.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service