Skip to Content
Merck
All Photos(1)

Key Documents

T1775

Sigma-Aldrich

L-Thyroxine

powder, BioReagent, suitable for cell culture

Synonym(s):

3,3′,5,5″-Tetraiodo-L-thyronine, 3-[4-(4-Hydroxy-3,5-diiodophenoxy)-3,5-diiodophenyl]-L-alanine, T4

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC6H2(I)2OC6H2(I)2CH2CH(NH2)CO2H
CAS Number:
Molecular Weight:
776.87
Beilstein:
2228515
EC Number:
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32190102
PubChem Substance ID:
NACRES:
NA.77
biological source:
synthetic
form:
powder
Assay:
≥98% (HPLC)

biological source

synthetic

Quality Level

product line

BioReagent

Assay

≥98% (HPLC)

form

powder

potency

5-50 ng/mL

technique(s)

cell culture | mammalian: suitable

mp

223 °C (dec.) (lit.)

solubility

4 M NH4OH in methanol: 50 mg/mL, clear to hazy, yellow

shipped in

ambient

storage temp.

room temp

SMILES string

N[C@@H](Cc1cc(I)c(Oc2cc(I)c(O)c(I)c2)c(I)c1)C(O)=O

InChI

1S/C15H11I4NO4/c16-8-4-7(5-9(17)13(8)21)24-14-10(18)1-6(2-11(14)19)3-12(20)15(22)23/h1-2,4-5,12,21H,3,20H2,(H,22,23)/t12-/m0/s1

InChI key

XUIIKFGFIJCVMT-LBPRGKRZSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

Thyroxine (T4) is a tetraiodo derivative of the p-hydroxyphenyl ether of tyrosine, which functions as growth factors or cell growth inhibitors. T4 is used to treat hypothyroidism and thyroid carcinoma. It regulates energy metabolism.

Application

L-Thyroxine has been used to investigate its role in colorectal cancer cell lines HCT 116 (APC wild type) and HT-29 (APC mutant). It has also been used to evaluate its effects on pro-inflammatory tumor necrosis factor-α (TNF-α), interleukin-6 (IL-6), angiogenic vascular endothelial growth factor (VEGF), leukocyte adhesion, intercellular adhesion molecule 1 (ICAM-1) and E-selectin in human umbilical vein endothelial cells (HUVECs).

Biochem/physiol Actions

L-Thyroxine (T4) and triiodo-L-thyronine (T3) are iodine-containing hormones produced from thyroglobulin in the thyroid follicular cells. The stimulation of metabolic rate and regulation of growth and development by these hormones appear to be due to their effects on DNA transcription and, thus, protein synthesis.

Preparation Note

For long term storage, keep stock solutions at -20° in working aliquots and avoid repeated freeze/thaw cycles. Solutions stored at 2-8 °C are stable for up to 30 days.

Reconstitution

To prepare 20 μg/ml stock solution, dissolve in 50 ml sterile culture medium per mg thyroxine.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

STOT RE 1

Target Organs

Thyroid,Cardio-vascular system,Kidney

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thyroid hormone promotes beta-catenin activation and cell proliferation in colorectal cancer
Lee YS, et al.
Hormones & Cancer, 9(3), 156-165 (2018)
The
Clinical Chemistry, 331(3), 174-180 (2008)
Thyroxine therapy
Toft AD
The New England Journal of Medicine, 331(3), 174-180 (1994)
Thyroid hormones (T3 and T4): dual effect on human cancer cell proliferation
Moriggi G, et al.
Anticancer Research, 31(1), 89-96 (2011)
Chemistry of thyroxine. III. Constitution and synthesis of thyroxine
Harrington CR and Barger G
The Biochemical Journal, 21(6), 169-183 (1927)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service