Skip to Content
Merck
All Photos(1)

Key Documents

S-106

Supelco

JWH-018 5-Pentanoic acid beta-D-glucuronide solution

100 μg/mL (Acetonitrile:Water (90:10) (v/v)), certified reference material, ampule of 1 mL

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H29NO9
CAS Number:
Molecular Weight:
547.55
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL (Acetonitrile:Water (90:10) (v/v))

application(s)

forensics and toxicology
forensics and toxicology

format

single component solution

storage temp.

−70°C

General description

The glucuronide metabolite of a secondary urinary metabolite of JWH-018, a synthetic cannabinoid found in several versions of the herbal mixture "Spice". Illicit use of this Spice cannabinoid centers on its psychoactive effects which mimic those of Δ9-THC in cannabis. This certified Snap-N-Spike® solution is suitable as starting material for calibrators and controls in Spice cannabinoid testing methods by GC/MS or LC/MS for urine drug testing, forensic analysis, and clinical toxicology applications.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michael B Manookin et al.
Neuron, 97(6), 1327-1340 (2018-03-06)
Considerable theoretical and experimental effort has been dedicated to understanding how neural circuits detect visual motion. In primates, much is known about the cortical circuits that contribute to motion processing, but the role of the retina in this fundamental neural
C Alex Goddard et al.
Journal of neurophysiology, 98(6), 3486-3493 (2007-09-28)
Cholinergic neurons in the parabigeminal nucleus of the rat midbrain were studied in an acute slice preparation. Spontaneous, regular action potentials were observed both with cell-attached patch recordings as well as with whole cell current-clamp recordings. The spontaneous activity of
Ping Liu et al.
Nature communications, 4, 1911-1911 (2013-05-30)
Nematode neurons generally produce graded potentials instead of action potentials. It is unclear how the graded potentials control postsynaptic cells under physiological conditions. Here we show that postsynaptic currents frequently occur in bursts at the neuromuscular junction of Caenorhabditis elegans.
Zoé Christenson Wick et al.
eLife, 8 (2019-10-15)
The hippocampus, a brain region that is important for spatial navigation and episodic memory, benefits from a rich diversity of neuronal cell-types. Through the use of an intersectional genetic viral vector approach in mice, we report novel hippocampal neurons which
José Luis Pérez-López et al.
Frontiers in neural circuits, 12, 70-70 (2018-09-14)
The dopaminergic neurons of the ventral tegmental area (VTA) have been identified with the ability to co-release dopamine and glutamate. This ability was first documented in the nucleus accumbens but showed to be absent in the dorsal striatum. Recently the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service