S-054
JWH-018 5-Hydroxypentyl metabolite solution
100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
About This Item
grade
certified reference material
Quality Level
form
liquid
feature
(Snap-N-Spike®)
packaging
ampule of 1 mL
manufacturer/tradename
Cerilliant®
drug control
Narcotic Licence Schedule E (Switzerland)
concentration
100 μg/mL in methanol
technique(s)
gas chromatography (GC): suitable
liquid chromatography (LC): suitable
application(s)
forensics and toxicology
forensics and toxicology
format
single component solution
storage temp.
−20°C
SMILES string
O=C(C1=CN(CCCCCO)C2=C1C=CC=C2)C3=CC=CC4=C3C=CC=C4
InChI
1S/C24H23NO2/c26-16-7-1-6-15-25-17-22(20-12-4-5-14-23(20)25)24(27)21-13-8-10-18-9-2-3-11-19(18)21/h2-5,8-14,17,26H,1,6-7,15-16H2
InChI key
UGHBAICDASCCIQ-UHFFFAOYSA-N
General description
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1
Target Organs
Eyes,Central nervous system
Storage Class Code
3 - Flammable liquids
WGK
WGK 2
Flash Point(F)
49.5 °F
Flash Point(C)
9.7 °C
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service