Skip to Content
Merck
All Photos(1)

Key Documents

O-013

Supelco

2-Oxo-3-hydroxy-LSD solution

100 μg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®

Synonym(s):

2-Oxo-3-hydroxy-LSD solution, 2-Oxo-3-hydroxy-lysergic acid diethylamide, 2-Oxo-3-hydroxy-lysergic acid diethylamide solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H25N3O3
CAS Number:
Molecular Weight:
355.43
EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL in acetonitrile

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

SMILES string

CCN(CC)C(=O)[C@H]1CN(C)[C@@H]2CC3(O)C(=O)Nc4cccc(C2=C1)c34

InChI

1S/C20H25N3O3/c1-4-23(5-2)18(24)12-9-14-13-7-6-8-15-17(13)20(26,19(25)21-15)10-16(14)22(3)11-12/h6-9,12,16,26H,4-5,10-11H2,1-3H3,(H,21,25)/t12-,16-,20?/m1/s1

InChI key

YSZSHHCNLVHCNV-VRORWYBRSA-N

General description

2-Oxo-3-hydroxy-LSD is a major urinary metabolite of lysergic acid diethyl amide, or LSD. This certified solution standard is suitable for use in LC/MS or GC/MS applications for clinical toxicology, forensic analysis, or urine drug testing. Known for its role in 1960′s American counterculture, LSD is one of the most potent psychedelic drugs ever created.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service