Skip to Content
Merck
All Photos(1)

Key Documents

H-178

Supelco

3-Hydroxybutyric acid-13C4 sodium salt solution

100 μg/mL in methanol (as free carboxylate), certified reference material, ampule of 1 mL, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13C4H7O3 · Na
CAS Number:
Molecular Weight:
130.06
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

Pricing and availability is not currently available.

grade

certified reference material

Quality Level

description

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL in methanol (as free carboxylate)

format

single component solution

storage temp.

-10 to -25°C

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
AXYVT38470ULRSAXYVT3845ULRAXYVT38470ULR
sterility

sterile

sterility

sterile

sterility

-

sterility

-

packaging

pkg of 5x3840tips/cs

packaging

pkg of 5x3840tips/cs

packaging

pkg of 5x3840tips/cs

packaging

pkg of 5x3840tips/cs

size

10 μL

size

70 μL

size

5 μL

size

70 μL

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service