Skip to Content
Merck
All Photos(1)

Key Documents

H-051

Supelco

Hydromorphone-3-β-D-glucuronide

100 μg/mL in methanol: water (1:1), ampule of 1 mL, certified reference material, Cerilliant®

Synonym(s):

Hydromorphone-3-beta-D-glucuronide solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H27NO9
CAS Number:
Molecular Weight:
461.46
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule A (Switzerland); estupefaciente (Spain); Decreto Lei 15/93: Tabela IA (Portugal)

concentration

100 μg/mL in methanol: water (1:1)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

2-8°C

SMILES string

CN1CC[C@@]23[C@H]4CCC(=O)[C@@H]2Oc5c(O[C@@H]6O[C@@H]([C@@H](O)[C@H](O)[C@H]6O)C(O)=O)ccc(C[C@@H]14)c35

InChI

1S/C23H27NO9/c1-24-7-6-23-10-3-4-12(25)20(23)32-18-13(5-2-9(14(18)23)8-11(10)24)31-22-17(28)15(26)16(27)19(33-22)21(29)30/h2,5,10-11,15-17,19-20,22,26-28H,3-4,6-8H2,1H3,(H,29,30)/t10-,11+,15-,16-,17+,19-,20-,22+,23-/m0/s1

InChI key

VLSHIBYUEOPPRX-NSSRKTJTSA-N

General description

Hydromorphone-3ß-D-glucuronide is a primary urinary metabolite of the opioid analgesic hydromorphone. This Certified Snap-N-Spike® Solution is suitable for use in LC/MS or GC/MS applications for forensic analysis, clinical toxicology, pain prescription monitoring or urine drug testing. Hydromorphone, marketed under brand names including Palladone and Dilaudid®, is a very potent opiate used as an analgesic for treating pain or as a cough suppressant for painful coughing resulting from continued bronchial irritation. 

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Dilaudid is a registered trademark of Purdue Pharma L.P.
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 3 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

75.2 °F - closed cup

Flash Point(C)

24 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

Optimize β-glucuronidase hydrolysis for glucuronide metabolite analysis considering factors like time, temperature, pH, and enzyme concentration.

Optimize β-glucuronidase hydrolysis for glucuronide metabolite analysis considering factors like time, temperature, pH, and enzyme concentration.

Optimize β-glucuronidase hydrolysis for glucuronide metabolite analysis considering factors like time, temperature, pH, and enzyme concentration.

Optimize β-glucuronidase hydrolysis for glucuronide metabolite analysis considering factors like time, temperature, pH, and enzyme concentration.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service