Skip to Content
Merck
All Photos(1)

Key Documents

E-004

Supelco

Ecgonine hydrochloride solution

1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H15NO3·HCl
CAS Number:
Molecular Weight:
221.68
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule A (Switzerland); estupefaciente (Spain); Decreto Lei 15/93: Tabela IB (Portugal)

concentration

1.0 mg/mL in methanol (as free base)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

2-8°C

SMILES string

Cl.CN1[C@H]2CC[C@@H]1[C@H]([C@@H](O)C2)C(O)=O

InChI

1S/C9H15NO3.ClH/c1-10-5-2-3-6(10)8(9(12)13)7(11)4-5;/h5-8,11H,2-4H2,1H3,(H,12,13);1H/t5-,6+,7-,8+;/m0./s1

InChI key

HVWQTEPEBQYIFB-PXXJPSRFSA-N

General description

Ecgonine is not only a precursor of cocaine, but also a metabolite obtained by cocaine hydrolysis. This analytical reference standard is suitable for use as starting material in calibrators and controls for a variety of LC/MS or GC/MS applications in cocaine testing from forensic analysis and clinical toxicology to urine drug testing.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M D Schreiber et al.
Journal of applied physiology (Bethesda, Md. : 1985), 77(2), 834-839 (1994-08-01)
Prenatal cocaine exposure has been reported to cause neurovascular complications in the developing fetus. To determine the effect of cocaine on the fetal neurovasculature, we studied the in vitro response of fetal sheep cerebral arteries to cocaine and cocaine metabolites.
Thomas Visalli et al.
Toxicology letters, 155(1), 35-40 (2004-12-09)
Hepatocellular damage is thought to occur as a result of cytochrome P450-mediated oxidation of cocaine to norcocaine (NC), a precursor of the hepatotoxic nitrosonium ion. However, this damage occurs only in male mice, with females exhibiting minimal biochemical and histological
Alexander L N van Nuijs et al.
Addiction (Abingdon, England), 104(5), 734-741 (2009-04-07)
Cocaine is the second most-used illicit drug world-wide and its consumption is increasing significantly, especially in western Europe. Until now, the annual prevalence has been estimated indirectly by means of interviews. A recently introduced and direct nation-wide approach based on
Matthew R Wood et al.
Acta crystallographica. Section C, Crystal structure communications, 66(Pt 1), m4-m8 (2010-01-06)
The structure of the hydrated gold(III) tetrachloride salt of L-ecgonine {hydronium tetrakis[(1R,2R,3S,5S,8S)-3-hydroxy-8-methyl-8-azoniabicyclo[3.2.1]octane-2-carboxylate pentakis[tetrachloridoaurate(III)] hexahydrate}, (C(9)H(16)NO(3))(4)(H(3)O)[AuCl(4)](5).6H(2)O, demonstrates an unprecedented stoichiometric relationship between the cations and anions in the unit cell. The previous tropane alkaloid structures, including the related hydrochloride salts, all
D L Lister et al.
Applied and environmental microbiology, 62(1), 94-99 (1996-01-01)
A mixed culture that could utilize cocaine as the sole source of carbon and energy for growth was isolated by selective enrichment. The individual microorganisms within this mixed culture were identified as Pseudomonas fluorescens (termed MBER) and Comamonas acidovorans (termed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service