Skip to Content
Merck
All Photos(1)

Key Documents

D-013

Supelco

Dextromethorphan solution

1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H25NO
CAS Number:
Molecular Weight:
271.40
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

SMILES string

COc1ccc2C[C@H]3[C@H]4CCCC[C@@]4(CCN3C)c2c1

InChI

1S/C18H25NO/c1-19-10-9-18-8-4-3-5-15(18)17(19)11-13-6-7-14(20-2)12-16(13)18/h6-7,12,15,17H,3-5,8-11H2,1-2H3/t15-,17+,18+/m1/s1

InChI key

MKXZASYAUGDDCJ-NJAFHUGGSA-N

Gene Information

General description

Dextromethorphan is an opiate analgesic sold as an antitussive under trade names such as Robitussin®, NyQuil®, or Dimetapp®. Also known as DXM, the drug is used recreationally as a hallucinogen with effects similar to ketamine and phencyclidine (PCP). This Certified Snap-N-Spike® Solution is suitable for use as starting material in calibrators or controls for a variety of LC/MS or GC/MS applications from forensic analysis and clinical toxicology to P450 trials and pharmaceutical research.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Dimetapp is a registered trademark of Wyeth LLC
NyQuil is a registered trademark of Procter & Gamble Co.
Robitussin is a registered trademark of Wyeth LLC
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

51.8 °F - closed cup

Flash Point(C)

11.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G Nicholas Verne et al.
The journal of pain : official journal of the American Pain Society, 13(9), 901-909 (2012-09-11)
Irritable bowel syndrome (IBS) is a common gastrointestinal disorder in which the pathophysiological mechanisms of the pain and hypersensitivity are incompletely understood. IBS patients frequently complain of pain in body regions somatotopically distinct from the gut, suggesting involvement of central
Andreas Trojan et al.
Cancer chemotherapy and pharmacology, 71(2), 301-306 (2012-10-27)
Endoxifen serum concentrations seem to correlate with outcome in breast cancer (BC) patients. Concurrently, cytochrome P450 2D6 (CYP2D6) enzyme activity and dextromethorphan (DM) metabolism are deemed a surrogate marker for the formation of endoxifen. Here, we conducted a matched cohort
Shiou-Lan Chen et al.
Journal of neuroimmune pharmacology : the official journal of the Society on NeuroImmune Pharmacology, 7(4), 1025-1033 (2012-09-20)
Recent studies show that proinflammatory cytokines might be related to the development of opioid dependence (physiological, psychological, or both). In a double-blind, randomly stratified clinical trial investigating whether add-on dextromethorphan (60-120 mg/day) attenuated inflammation and the combined use of opioids
Shinn-Jong Jiang et al.
Microcirculation (New York, N.Y. : 1994), 20(2), 190-201 (2012-11-13)
This study examines the effect of Dextromethorphan (d-3-methoxy-17-methylmorphinan; DXM), a commonly used cough-suppressing drug, on the expression of VCAM-1 and ICAM-1 in human umbilical vein endothelial cells (HUVECs) stimulated with lipopolysaccharide (LPS). The effect of DXM on expression of cell
F L Opdam et al.
Cancer chemotherapy and pharmacology, 71(3), 593-601 (2012-12-12)
Adjuvant therapy with tamoxifen significantly reduces breast cancer recurrence and mortality in estrogen receptor positive disease. CYP2D6 is the main enzyme involved in the activation of the prodrug tamoxifen into the anti-estrogen endoxifen. Endoxifen is thought to be a main

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service