Skip to Content
Merck
All Photos(1)

Key Documents

878112C

Avanti

C16-18:1 PC

Avanti Research - A Croda Brand 878112C

Synonym(s):

1-O-hexadecyl-2-oleoyl-sn-glycero-3-phosphocholine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C42H84NO7P
CAS Number:
Molecular Weight:
746.09
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

form

liquid

packaging

pkg of 1 × 1 mL (878112C-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 878112C

concentration

5 mg/mL (878112C-5mg)

lipid type

phosphoglycerides
bioactive lipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[O-]P(OCC[N+](C)(C)C)(OC[C@]([H])(OC(CCCCCCC/C=C\CCCCCCCC)=O)COCCCCCCCCCCCCCCCC)=O

InChI key

SIEDNCDNGMIKST-IYEJTHTFSA-N

General description

C16-18:1 PC, also referred as 1-O-hexadecyl-2-oleoyl-sn-glycero-3-phosphocholine (HOPC), is a platelet activating factor. It belongs to the class of bioactive lipids. HOPC is a mono-ether PC containing ester-linked 18:1 chain (oleoyl) at the sn-2 position and an ether-linked 16:0 chain (hexadecyl) at the sn-1 position.

Application

C16-18:1 PC or 1-O-hexadecyl-2-oleoyl-sn-glycero-3-phosphocholine may be used: as a lipid standard for quantification of glycerophosphocholine by liquid chromatography–mass spectrometry (LC–MS), in preparation of phospholipid vesicles, as a substrate to study the substrate specificity of phospholipase B/lipase (PLB/LIP) in the presence and absence of deoxycholate (DOC).

Biochem/physiol Actions

Platelet-activating factor (PAF) acts as a lipid mediator and is involved in various inflammatory diseases.

Packaging

5 mL Clear Glass Sealed Ampule (878112C-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Matts Nylund et al.
Biochimica et biophysica acta, 1758(6), 807-812 (2006-06-17)
The glycolipid transfer protein (GLTP)-mediated movement of galactosylceramide from model membrane donor vesicles to acceptor vesicles is sensitive to the membrane environment surrounding the glycolipid. GLTP can catalyze the transfer of a fluorescently labeled GSL, anthrylvinyl-galactosylceramide (AV-GalCer), from vesicles composed
H Tojo et al.
The Journal of biological chemistry, 273(4), 2214-2221 (1998-01-27)
A brush border membrane-associated phospholipase B/lipase was solubilized from the distal two-thirds of rat small intestine by autolysis during storage at -35 degrees C over 1 month, and then the enzyme was purified to homogeneity and characterized enzymatically and structurally.
Akira Sato et al.
European journal of pharmacology, 745, 46-51 (2014-12-03)
Platelet-activating factor (PAF) is a potent lipid mediator that is implicated in numerous inflammatory diseases. C-reactive protein (CRP) is an acute-phase plasma protein that increases rapidly and dramatically in response to inflammation. In this study, we investigated the effect of
Ernst R Werner et al.
Proceedings of the National Academy of Sciences of the United States of America, 117(14), 7792-7798 (2020-03-27)
A significant fraction of the glycerophospholipids in the human body is composed of plasmalogens, particularly in the brain, cardiac, and immune cell membranes. A decline in these lipids has been observed in such diseases as Alzheimer's and chronic obstructive pulmonary
Andrey B Imbs et al.
PloS one, 14(4), e0215759-e0215759 (2019-04-30)
To expand our knowledge of lipid and fatty acid (FA) biosynthesis in marine cnidarians, polar lipidomes of hydrocorals were studied for the first time and then compared with those of soft corals from tropical and boreal regions. The structure and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service