Skip to Content
Merck
All Photos(2)

Key Documents

852758C

Avanti

C18(Plasm)-18:1 PE

Avanti Research - A Croda Brand

Synonym(s):

1-O-1′-(Z)-octadecenyl-2--(9Z-octadecenoyl)-sn-glycero-3-phosphoethanolamine; GPEtn(18:0p/18:1(9Z)); 110645

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C41H80NO7P
CAS Number:
Molecular Weight:
730.05
UNSPSC Code:
51191904
NACRES:
NA.25
Pricing and availability is not currently available.

description

1-(1Z-octadecenyl)-2-oleoyl-sn-glycero-3-phosphoethanolamine, chloroform

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 5 mL (852758C-10mg)
pkg of 1 × 5 mL (852758C-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand

concentration

1 mg/mL (852758C-5mg)
2 mg/mL (852758C-10mg)

lipid type

phospholipids
cardiolipins

shipped in

dry ice

storage temp.

−20°C

SMILES string

[O-]P(OCC[NH3+])(OC[C@]([H])(OC(CCCCCCC/C=C\CCCCCCCC)=O)CO/C=C\CCCCCCCCCCCCCCCC)=O

InChI key

XVYPOHCSLJZFED-QZEVRULJSA-N

General description

Phosphatidylethanolamine is an aminophospholipid. It is present in eukaryotic and prokaryotic cells.[1] Phosphatidylethanolamine is one of the most abundant glycerophospholipids in the biological membranes.[2] It is found in nervous tissue.[3]

Application

C18(Plasm)-18:1 PE has been used as a lipid standard in the optimization of the liquid chromatography-mass spectrometry (LC-MS).[4]

Biochem/physiol Actions

Phosphatidylethanolamine forms pure lipid structures,[5] which regulate membrane fluidity in eukaryotic cells.[6] Phosphatidylethanolamine plays a major role in autophagy, cell division and protein folding. It functions as a precursor for the synthesis of various protein modifications. Phosphatidylethanolamine acts as an intermediate for the synthesis of glycerophospholipid classes.[2]

Packaging

30 mL Amber Narrow Mouth Glass Bottle with Screw Cap (852758C-10mg)
30 mL Amber Narrow Mouth Glass Bottle with Screw Cap (852758C-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

also commonly purchased with this product

Product No.
Description
Pricing

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Oral - STOT SE 3

Target Organs

Liver,Kidney, Respiratory system

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

An Introduction to Biological Membranes: Composition, Structure and Function (2016)
The Membranes of Cells (2016)
Nathan R Deleault et al.
Proceedings of the National Academy of Sciences of the United States of America, 109(28), E1938-E1946 (2012-06-20)
Prions containing misfolded prion protein (PrP(Sc)) can be formed with cofactor molecules using the technique of serial protein misfolding cyclic amplification. However, it remains unknown whether cofactors materially participate in maintaining prion conformation and infectious properties. Here we show that
Ana West et al.
The journal of physical chemistry. B, 124(5), 828-839 (2020-01-10)
Ethanolamine plasmalogen (EtnPLA) is a conical-shaped ether lipid and an essential component of neurological membranes. Low stability against oxidation limits its study in experiments. The concentration of EtnPLA in the bilayer varies depending on cell type and disease progression. Here
Geoffrey P Noble et al.
Biochemistry, 54(5), 1180-1187 (2015-01-15)
Misfolding of the prion protein (PrP) plays a central role in the pathogenesis of infectious, sporadic, and inherited prion diseases. Here we use a chemically defined prion propagation system to study misfolding of the pathogenic PrP mutant D177N in vitro.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service