Skip to Content
Merck
All Photos(1)

Key Documents

860599C

Avanti

C18:1 Ceramide-1-Phosphate (d18:1/18:1(9Z))

Avanti Research - A Croda Brand 860599C

Synonym(s):

N-oleoyl-ceramide-1-phosphate (ammonium salt)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C36H73N2O6P
CAS Number:
Molecular Weight:
660.95
UNSPSC Code:
12352211
NACRES:
NA.25

form

liquid

packaging

pkg of 1 × 1 mL (860599C-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 860599C

concentration

1 mg/mL (860599C-1mg)

lipid type

sphingolipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@](/C=C/CCCCCCCCCCCCC)(O)[C@@]([H])(NC(CCCCCCC/C=C\CCCCCCCC)=O)COP([O-])(O)=O.[NH4+]

InChI key

OLFFCEGYMCQDKZ-CFRDMXQMSA-N

General description

Ceramide-1-phosphate (C1P) is a ceramide-derived metabolite, which belongs to the class of bioactive lipid mediators.

Application

C18:1 Ceramide-1-Phosphate (d18:1/18:1(9Z)) may be used for fluorescence analysis of phosphatidic acid-binding tetravalent peptide (PAB-TP) phospholipid specificity. It may also be used in liposome cosedimentation assay to compare its affinity with α-synuclein-N.

Biochem/physiol Actions

Ceramide-1-phosphate (C1P) regulates cell survival, cell growth and death. It mediates inflammatory response and phagocytosis.

Packaging

5 mL Clear Glass Sealed Ampule (860599C-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Characterization of alpha-synuclein N-terminal domain as a novel cellular phosphatidic acid sensor
Yamada H, et al.
The FEBS journal (2019)
Characterization of alpha-synuclein N-terminal domain as a novel cellular phosphatidic acid sensor
Yamada H, et al.
FEBS Journal (2019)
Development of a novel tetravalent synthetic peptide that binds to phosphatidic acid
Ogawa R, et al.
Testing, 10(7) (2015)
Blood sphingolipidomics in healthy humans: impact of sample collection methodology
Hammad SM, et al.
Journal of Lipid Research, 51(10), 3074-3087 (2010)
Development of a novel tetravalent synthetic peptide that binds to phosphatidic acid
Ogawa R, et al.
PLoS ONE, 10(7) (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service