Skip to Content
Merck
All Photos(2)

Key Documents

860053P

Avanti

Ganglioside-Total

Avanti Research - A Croda Brand

Synonym(s):

Total Ganglioside Extract (Brain, Porcine-Ammonium Salt)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 10 mg (860053P-10mg)
pkg of 1 × 25 mg (860053P-25mg)
pkg of 1 × 50 mg (860053P-50mg)

manufacturer/tradename

Avanti Research - A Croda Brand

lipid type

sphingolipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@](/C=C/CCCCCCCCCCCCC)(O)[C@@]([H])(NC(CCCCCCCCCCCCCCCCC)=O)CO[C@H]1[C@H](O)[C@@H](O)[C@H](O[C@@H]2O[C@H](CO)[C@@H]([C@H](O[C@@]3(C([O-])=O)O[C@@H]([C@H](O)[C@H](O)CO)[C@@](NC(C)=O)([H])[C@@H](O)C3)[C@H]2O)O[C@@H]4O[C@H](CO)[C@@H]([C@H](O[C@@H]5O[C@H

General description

Gangliosides are glycosphingolipids, that contain sialic acid residues in the glycan chain. They are mainly present on the surface of neuronal cells. Gangliosides are also present in serum. Four complex ganglioside structures, such as GM1, GD1a, GD1b and GT1b are the most abundantly found ganglioside in mammalian brains. Gangliosides comprise a ceramide tail attached to a polar carbohydrate head.

Application

Ganglioside-Total may be used:
  • as a standard to investigate the efficiency of ganglioside extraction in brain using electrospray ionization-mass spectrometry (ESI MS) analysis
  • as a hemagglutinin (HA) binding receptor to assemble nanodisc with affinity to influenza virus (NDTG) and also to determine the effect of total ganglioside extract (TG) amount on total ganglioside
  • in liposomes to measure ostreolysin A (OlyA) binding in order to deduce the effects of phosphatidylcholines (PCs) on sphingomyelin

Biochem/physiol Actions

Gangliosides regulate various biological activities in cell plasma membranes (PMs). It provides structural rigidity to the cell. Ganglioside clusters form membrane microdomains, which facilitates cell adhesion. It also functions as differentiating markers during oncogenesis, embryogenesis, lymphoid differentiation and neoplastic transformation. Gangliosides have the ability to repress cell-mediated immune functions. It is also associated with cancer and Alzheimer′s disease.

Packaging

5 mL Amber Glass Screw Cap Vial (860053P-10mg)
5 mL Amber Glass Screw Cap Vial (860053P-25mg)
5 mL Amber Glass Screw Cap Vial (860053P-50mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

No data available

Flash Point(C)

No data available


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Byung-Hwan Lee et al.
Neuroscience letters, 612, 256-260 (2015-12-27)
Ginseng has a long history of use as a tonic for restoration of vigor. One example of ginseng-derived tonic effect is that it can improve physical stamina under conditions of stress. However, the active ingredient and the underlying molecular mechanism
Jin Liu et al.
Biophysical journal, 89(4), 2522-2532 (2005-08-09)
Sum-frequency vibrational spectroscopy (SFVS) is used to measure the intrinsic rate of lipid flip-flop for 1,2-dimyristoyl-sn-glycero-3-phosphocholine (DMPC), 1,2-dipalmitoyl-sn-glycero-3-phosphocholine (DPPC), and 1,2-distearoyl-sn-glycero-3-phosphocholine (DSPC) in planar-supported lipid bilayers (PSs). Asymmetric PSLBs were prepared using the Langmuir-Blodgett/Langmuir-Schaefer method by placing a perdeuterated lipid
Raphael Dos Santos Morais et al.
Biophysical journal, 115(7), 1231-1239 (2018-09-11)
Scaffolding proteins play important roles in supporting the plasma membrane (sarcolemma) of muscle cells. Among them, dystrophin strengthens the sarcolemma through protein-lipid interactions, and its absence due to gene mutations leads to the severe Duchenne muscular dystrophy. Most of the
Thomas Kolter
ISRN biochemistry, 2012, 506160-506160 (2012-01-01)
Gangliosides are sialic acid-containing glycosphingolipids. They occur especially on the cellular surfaces of neuronal cells, where they form a complex pattern, but are also found in many other cell types. The paper provides a general overview on their structures, occurrence
Petru Niga et al.
Journal of colloid and interface science, 526, 230-243 (2018-05-08)
Vibrational sum frequency spectroscopy (VSFS) complemented by surface pressure isotherm and neutron reflectometry (NR) experiments were employed to investigate the interactions between propofol, a small amphiphilic molecule that currently is the most common general anaesthetic drug, and phospholipid monolayers. A

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service