Merck
Ganglioside biochemistry.
ISRN biochemistry (2012-01-01)
Thomas Kolter
ABSTRACT
Gangliosides are sialic acid-containing glycosphingolipids. They occur especially on the cellular surfaces of neuronal cells, where they form a complex pattern, but are also found in many other cell types. The paper provides a general overview on their structures, occurrence, and metabolism. Key functional, biochemical, and pathobiochemical aspects are summarized.
Product Description
Avanti
C18:0 GM3 (synthetic), Avanti Research - A Croda Brand 860074W, methanol solution
Avanti
Sphingosine (d20:1), Avanti Research - A Croda Brand 860660P, powder
Avanti
Ganglioside GM3 (Bovine Milk), Avanti Research - A Croda Brand