Skip to Content
Merck
All Photos(2)

Key Documents

850187P

Avanti

08:0 PI(3)P

1,2-dioctanoyl-sn-glycero-3-(phosphoinositol-3-phosphate) (ammonium salt), powder

Synonym(s):

1,2-dioctanoyl-sn-glycero-3-phospho-(1′-myo-inositol-3′-phosphate) (ammonium salt); PIP[3′](8:0/8:0)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H54N2O16P2
CAS Number:
Molecular Weight:
700.65
UNSPSC Code:
51191904
NACRES:
NA.25

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 100 μg (with stopper and crimp cap (850187P-100ug))
pkg of 1 × 500 μg (with stopper and crimp cap (850187P-500ug))

manufacturer/tradename

Avanti Research - A Croda Brand 850187P

lipid type

phospholipids
cardiolipins

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(O[C@H]1[C@H](O)[C@@H](O)[C@H](O)[C@@H](OP([O-])(O)=O)[C@H]1O)=O)(OC(CCCCCCC)=O)COC(CCCCCCC)=O.[NH4+].[NH4+]

InChI

1S/C25H48O16P2.2H3N/c1-3-5-7-9-11-13-18(26)37-15-17(39-19(27)14-12-10-8-6-4-2)16-38-43(35,36)41-25-22(30)20(28)21(29)24(23(25)31)40-42(32,33)34;;/h17,20-25,28-31H,3-16H2,1-2H3,(H,35,36)(H2,32,33,34);2*1H3/t17-,20+,21+,22-,23-,24-,25+;;/m1../s1

InChI key

IHRPLAYZBAFIEF-ACHOKTOGSA-N

General description

Phosphatidylinositol 3-monophosphate (PI3P) is a lipid, found abundantly in eukaryotic cells. It is also present at low level in mammalian cells.

Application

08:0 PI(3)P has been used in the preparation of model membrane for protein reconstitution and structural studies.

Biochem/physiol Actions

Phosphatidylinositol 3-monophosphate (PI3P) participates in vesicular trafficking processes. It also plays a key role in endocytosis.

Packaging

2 mL Amber Serum Vial with Stopper and Crimp Cap (850187P-100ug)
2 mL Amber Serum Vial with Stopper and Crimp Cap (850187P-500ug)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lina Tawk et al.
Eukaryotic cell, 9(10), 1519-1530 (2010-08-17)
Phosphoinositides are important regulators of diverse cellular functions, and phosphatidylinositol 3-monophosphate (PI3P) is a key element in vesicular trafficking processes. During its intraerythrocytic development, the malaria parasite Plasmodium falciparum establishes a sophisticated but poorly characterized protein and lipid trafficking system.
Structural and functional studies of membrane remodeling machines
Methods in Cell Biology, 128, 165-200 (2015)
Etienne Morel et al.
Nature communications, 4, 2250-2250 (2013-08-03)
Defects in endosomal sorting have been implicated in Alzheimer's disease. Endosomal traffic is largely controlled by phosphatidylinositol-3-phosphate, a phosphoinositide synthesized primarily by lipid kinase Vps34. Here we show that phosphatidylinositol-3-phosphate is selectively deficient in brain tissue from humans with Alzheimer's

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service