Skip to Content
Merck
All Photos(4)

Key Documents

L5254

Sigma-Aldrich

1-Palmitoyl-sn-glycero-3-phosphocholine

synthetic, ≥99%

Synonym(s):

PC(16:0/0:0), 1-Hexadecanoyl-sn-glycero-3-phosphocholine, 3-sn-Lysophosphatidylcholine, 1-palmitoyl, L-α-Palmitoyl-lysophosphatidylcholine, L-γ-Palmitoyl-α-lysolecithin, Lysolecithin, palmitoyl

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H50NO7P
CAS Number:
Molecular Weight:
495.63
Beilstein:
5385682
MDL number:
UNSPSC Code:
51191904
PubChem Substance ID:
NACRES:
NA.25

biological source

synthetic

Quality Level

Assay

≥99%

form

powder

functional group

phospholipid

lipid type

phosphoglycerides

shipped in

ambient

storage temp.

−20°C

SMILES string

O[C@](COP([O-])(OCC[N+](C)(C)C)=O)([H])COC(CCCCCCCCCCCCCCC)=O

InChI

1S/C24H50NO7P/c1-5-6-7-8-9-10-11-12-13-14-15-16-17-18-24(27)30-21-23(26)22-32-33(28,29)31-20-19-25(2,3)4/h23,26H,5-22H2,1-4H3/t23-/m1/s1

InChI key

ASWBNKHCZGQVJV-HSZRJFAPSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

1-Palmitoyl-sn-glycero-3-phosphocholine may be used as a substrate to identify, differentiate and characterize lysophosphocholine acyl transferases.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nicolas Blondeau et al.
Journal of cerebral blood flow and metabolism : official journal of the International Society of Cerebral Blood Flow and Metabolism, 22(7), 821-834 (2002-07-27)
Lysophospholipids (LPLs) are important intermediates in the synthesis and degradation of membrane phospholipids. Here we show that certain LPLs, particularly lysophosphatidylcholine and lysophosphatidylinositol, prevent neuronal death both in an in vivo model of transient global ischemia and in an in
Inna A Nikonorova et al.
Nucleic acids research, 42(20), 12779-12788 (2014-10-19)
TRPM7 is an essential and ubiquitous channel-kinase regulating cellular influx of Mg2+. Although TRPM7 mRNA is highly abundant, very small amount of the protein is detected in cells, suggesting post-transcriptional regulation of trpm7 gene expression. We found that TRPM7 mRNA
Akira Sato et al.
Chemical biology & drug design, 80(3), 417-425 (2012-05-18)
Lysophosphatidylcholine, a major phospholipid component of oxidized low-density lipoprotein, is implicated in many inflammatory diseases, including atherosclerosis. We previously reported that Asp-hemolysin-related synthetic peptide (P21) composed of 21 amino acid residues markedly inhibits the bioactivities of oxidized low-density lipoprotein and
Masanori Iwase et al.
Atherosclerosis, 196(2), 931-936 (2007-03-14)
Increased lipoprotein-associated PLA(2) (Lp-PLA(2)) predicts the future development of cardiovascular diseases. Although lysophosphatidylcholine (lyso-PC) produced by Lp-PLA(2) may contribute to its proatherogenic activity, the relation between Lp-PLA(2) and lyso-PC content in LDL remains unclarified. We determined the correlation between lyso-PC
Lu-Cheng Cao et al.
Kidney international, 66(5), 1890-1900 (2004-10-22)
In cultured renal epithelial cells, exposure to oxalate, a constituent of many kidney stones, elicits a cascade of responses that often leads to cell death. Oxalate toxicity is mediated via generation of reactive oxygen species (ROS) in a process that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service